317497
Dimethyl 3,4-furandicarboxylate
98%
Synonym(s):
3,4-Bis(methoxycarbonyl)furan, 3,4-Dimethyl 3,4-furandicarboxylate, 3,4-Dimethyl furan-3,4-dicarboxylate
About This Item
Recommended Products
Assay
98%
form
solid
bp
91-93 °C/1 mmHg (lit.)
mp
46-49 °C (lit.)
functional group
ester
SMILES string
COC(=O)c1cocc1C(=O)OC
InChI
1S/C8H8O5/c1-11-7(9)5-3-13-4-6(5)8(10)12-2/h3-4H,1-2H3
InChI key
SEVRCIAIDGNMFY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Luminescent lanthanide-furandicarboxylate coordination polymer through hydrolysis with lanthanide ions.
- Polyesters via polycondensation with linear biobased aliphatic diols.
- Furanmonomers via lithium aluminium hydride (LiAlH4) reduction, which then react with maleimides to form furan-maleimide Diels-Alder adducts for thermally responsive dendrons and dendrimers.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service