Skip to Content
Merck
All Photos(2)

Key Documents

310905

Sigma-Aldrich

Diethylamine hydrobromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH · HBr
CAS Number:
Molecular Weight:
154.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

218-220 °C (lit.)

functional group

amine

SMILES string

Br[H].CCNCC

InChI

1S/C4H11N.BrH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1H

InChI key

AATGHKSFEUVOPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylamine hydrobromide was used in the synthesis of α-bromovinyltrimethylsilane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESES AND POLYMERIZATION OF α-TRIMETHYLSILYL ACRYLIC MONOMERS.
Canadian Journal of Chemistry, 41(12), 2977- 2982 (1963)
Zhao Chunfang et al.
Journal of chromatographic science, 48(8), 685-689 (2010-09-08)
The thin-layer chromatography (TLC) combined with high-performance liquid chromatography (HPLC) has been proved to be a quick and valid method to detect the intermediates and the end-product created during the chemosynthesis process of vinorelbine (VB). This paper gives a detailed
V M Gun'ko et al.
Journal of colloid and interface science, 348(2), 546-558 (2010-07-14)
Adsorption of low-molecular adsorbates (nonpolar hexane, nitrogen, weakly polar acetonitrile, and polar diethylamine, triethylamine, and water) onto individual (silica, alumina, titania), binary (silica/alumina (SA), silica/titania (ST)), and ternary (alumina/silica/titania, AST) fumed oxides was studied to analyse the effects of morphology
Chunhe Yao et al.
Journal of separation science, 33(4-5), 475-483 (2010-01-12)
A novel kind of poly(glycidyl methacrylate-co-ethylene glycol dimethacrylate)-based monolithic column was developed for LC by directing supramolecular self-assembly of high internal phase emulsion. Mercury intrusion porosimetry characterization and scanning electron microscope pictures showed that these monoliths presented micrometer-sized throughpores, unique
National Toxicology Program technical report series, (566)(566), 1-174 (2011-12-01)
Diethylamine is used mainly as a chemical intermediate to produce the corrosion inhibitor N,N-diethylethanolamine and a lesser amount is used to produce pesticides and insect repellants and in rubber processing. Diethylamine was nominated for study by the National Institute of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service