All Photos(2)
About This Item
Linear Formula:
CH3CH2CH2CO2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
6 mmHg ( 20 °C)
Assay
99%
form
liquid
refractive index
n20/D 1.393 (lit.)
bp
130-131 °C (lit.)
density
0.859 g/mL at 25 °C (lit.)
functional group
ester
SMILES string
CCCC(=O)OC(C)C
InChI
1S/C7H14O2/c1-4-5-7(8)9-6(2)3/h6H,4-5H2,1-3H3
InChI key
FFOPEPMHKILNIT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Isopropyl butyrate is an electroantennographic detection (EAD) active compound that was identified by GC-mass spectrometry. It is attractive to both sexes of West Indian fruit fly, Anastrepha obliqua (Macquart) (Diptera: Tephritidae).
Application
Isopropyl butyrate was used as intravaginal taggant in the study of "breath test" for vaginal gel use.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
75.2 °F
Flash Point(C)
24 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jorge Toledo et al.
Journal of economic entomology, 102(6), 2072-2077 (2010-01-15)
Previous studies have shown that a nine-component blend (ethyl butyrate, isopropyl butyrate, hexan-1-ol, propyl butyrate, isobutyl butyrate, ethyl hexanoate, isopentyl butyrate, ethyl benzoate, and ethyl octanoate) isolated from Spondias mombin L. (Anacardiaceae) fruit are attractive to both sexes of West
Leopoldo Cruz-López et al.
Journal of chemical ecology, 32(2), 351-365 (2006-03-24)
Nonirradiated males and females of Anastrepha obliqua (Macquart) were attracted to and landed more frequently on ripe fruits of Spondias mombin L. than on artificial fruit in wind tunnel bioassays. Porapak Q volatile extracts of S. mombin were also attractive
Timothy E Morey et al.
Journal of clinical pharmacology, 53(1), 103-111 (2013-02-13)
Adherence to microbicide gel use is critical to optimizing effectiveness in preventing human immunodeficiency virus transmission. The authors hypothesized that ester taggants added to vaginal gels would generate exhaled alcohol and ketone metabolites and provide a "breath test" for vaginal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service