Skip to Content
Merck
All Photos(2)

Key Documents

302880

Sigma-Aldrich

2-Methoxy-4-methylphenol

≥98%

Synonym(s):

2-Hydroxy-5-methylanisole, 2-Methoxy-p-cresol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, Creosol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1862340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

refractive index

n20/D 1.537 (lit.)

bp

221-222 °C (lit.)

mp

5 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

SMILES string

COc1cc(C)ccc1O

InChI

1S/C8H10O2/c1-6-3-4-7(9)8(5-6)10-2/h3-5,9H,1-2H3

InChI key

PETRWTHZSKVLRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxy-4-methylphenol is the major component of black-ripe table olive aroma. It is the major anti-inflammatory compound in bamboo vinegar. Kinetics of reaction of 2-methoxy-4-methylphenol with chlorine atoms was studied.

Application

2-Methoxy-4-methylphenol was used in preparation of renewable bis(cyanate) esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Lung Ho et al.
PloS one, 8(10), e75738-e75738 (2013-10-15)
Bamboo vinegar (BV), a natural liquid derived from the condensation produced during bamboo charcoal production, has been used in agriculture and as a food additive, but its application to immune modulation has not been reported. Here, we demonstrated that BV
Amélie Lauraguais et al.
The journal of physical chemistry. A, 118(10), 1777-1784 (2014-02-28)
The reaction of a series of oxygenated aromatics (two methoxybenzene and six methoxyphenol isomers) with chlorine atoms has been studied in two simulation chambers with volumes of 1080 and 480 L at the University of Wuppertal. Experiments were performed at
Angelina Sansone-Land et al.
Food chemistry, 149, 285-295 (2013-12-04)
Volatile constituents of commercial black-ripe table olives (Olea europaea) from the United States, Spain, Egypt and Morocco were analysed by gas chromatography and gas chromatography-mass spectrometry (GC-MS). Dynamic headspace sampling was used to isolate a variety of aldehydes, alcohols, esters
Heather A Meylemans et al.
Biomacromolecules, 14(3), 771-780 (2013-01-18)
A series of renewable bis(cyanate) esters have been prepared from bisphenols synthesized by condensation of 2-methoxy-4-methylphenol (creosol) with formaldehyde, acetaldehyde, and propionaldehyde. The cyanate esters have been fully characterized by infrared spectroscopy, (1)H and (13)C NMR spectroscopy, and single crystal
Ana-Lilia González-Yebra et al.
International archives of occupational and environmental health, 79(3), 259-264 (2005-07-08)
An important, although, unprecise number of shoe workers in Leon, Mexico, are in continuous contact with toluene-based glues. The induction of renal glomerular and/or tubular lesions as a result of toluene exposure is still being discussed controversially. Our objective was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service