Skip to Content
Merck
All Photos(1)

Key Documents

302864

Sigma-Aldrich

Tri(propylene glycol) methyl ether, mixture of isomers

96%

Synonym(s):

O-Methyltripropylene glycol, [2-(2-Methoxymethylethoxy)methylethoxy]propanol, methylpropylene triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OC3H6)3OH
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.43 (lit.)

bp

100 °C/2 mmHg (lit.)

density

0.968 g/mL at 25 °C

SMILES string

O(C(COC(COC)C)C)C(CO)C

InChI

1S/C10H22O4/c1-8(5-11)14-10(3)7-13-9(2)6-12-4/h8-11H,5-7H2,1-4H3

InChI key

GVZNXUAPPLHUOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tri(propylene glycol) methyl ether is a promising oxygenating agent for diesel fuels.[1] Bubble coalescence and post-rupture oscillation in tri(propylene glycol) methyl ether solutions using high-speed cinematography was studied.[2]

Application


  • Composition-explicit distillation curves of diesel fuel with glycol ether and glycol ester oxygenates: fuel analysis metrology to enable decreased particulate emissions.: This study by Smith, Ott, and Bruno explores the application of glycol ethers, including tri(propylene glycol) methyl ether, in diesel fuels. The research demonstrates how these oxygenates can decrease particulate emissions, providing a valuable insight into the environmental benefits and performance improvements of glycol ethers in fuel formulations (Smith et al., 2008).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Bournival et al.
Journal of colloid and interface science, 414, 50-58 (2013-11-16)
Most processes involving bubbling in a liquid require small bubbles to maximise mass/energy transfer. A common method to prevent bubbles from coalescing is by the addition of surfactants. In order to get an insight into the coalescence process, capillary bubbles
Beverly L Smith et al.
Environmental science & technology, 42(20), 7682-7689 (2008-11-06)
We recently introduced several important improvements in the measurement of distillation curves of complex fluids. The modifications to the classical measurement provide for (1) a composition-explicit data channel for each distillate fraction (for both qualitative and quantitative analysis), (2) temperature

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service