Skip to Content
Merck
All Photos(1)

Documents

29524

Sigma-Aldrich

(R)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis[bis(3,5-di-tert-butylphenyl)phosphine]

≥97%, optical purity ee: ≥99%

Synonym(s):

(R)-2,2′-Bis[bis(3,5-di-tert-butyl)phosphino]-6,6′-dimethoxy-1,1′-biphenyl, (R)-3,5-t-Bu-MeOBIPHEP, SL-A121-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H96O2P2
CAS Number:
Molecular Weight:
1031.46
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

optical purity

ee: ≥99%

SMILES string

COc1cccc(P(c2cc(cc(c2)C(C)(C)C)C(C)(C)C)c3cc(cc(c3)C(C)(C)C)C(C)(C)C)c1-c4c(OC)cccc4P(c5cc(cc(c5)C(C)(C)C)C(C)(C)C)c6cc(cc(c6)C(C)(C)C)C(C)(C)C

InChI

1S/C70H96O2P2/c1-63(2,3)45-33-46(64(4,5)6)38-53(37-45)73(54-39-47(65(7,8)9)34-48(40-54)66(10,11)12)59-31-27-29-57(71-25)61(59)62-58(72-26)30-28-32-60(62)74(55-41-49(67(13,14)15)35-50(42-55)68(16,17)18)56-43-51(69(19,20)21)36-52(44-56)70(22,23)24/h27-44H,1-26H3

InChI key

PBYRAYONARLAQJ-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Application

(R)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis[bis(3,5-di-tert-butylphenyl)phosphine] can be used as a ligand to prepare:
  • 1,2-Dihydropyridines through Rh-catalyzed cycloaddition of diynes to sulfonimines.
  • Enantiorich disubstituted γ-lactams via intramolecular allylic alkylation reaction using Pd catalyst.
  • A Ru-metal complex, which acts as a hydrogenation catalyst applicable in the synthesis of an organic building block (S)-3-fluoromethyl-γ-butyrolactone.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective γ-lactam synthesis via palladium-catalyzed intramolecular asymmetric allylic alkylation
Bantreil X, et al.
Synlett, 2009(09), 1441-1444 (2009)
Highly enantioselective rhodium-catalyzed [2+2+2] cycloaddition of diynes to sulfonimines
Amatore M, et al.
Journal of the American Chemical Society, 135(12), 4576-4579 (2013)
Development of a scalable synthesis of (S)-3-fluoromethyl-γ-butyrolactone, building block for Carmegliptin′s lactam moiety
Adam J-M, et al.
Organic Process Research & Development, 15(3), 515-526 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service