293938
Tetradecyl sulfate sodium salt
95%
Synonym(s):
Sodium myristyl sulfate, Sodium tetradecyl sulfate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3(CH2)13OSO3Na
CAS Number:
Molecular Weight:
316.43
Beilstein:
3578122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
SMILES string
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
InChI
1S/C14H30O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-18-19(15,16)17;/h2-14H2,1H3,(H,15,16,17);/q;+1/p-1
InChI key
UPUIQOIQVMNQAP-UHFFFAOYSA-M
General description
Tetradecyl sulfate sodium salt is a detergent-like sclerosant.
Application
Tetradecyl sulfate sodium salt (Sodium tetradecyl sulphate) has been used:
- to investigate the effects of sclerosant foam preparation and composition on foam structure, the time course of liquid drainage and foam coarsenin
- in sclerotherapy of telangiectases and reticular veins
- in situ studies of self-assembled monolayers of adsorbed cationic or anionic surfactant molecules on atomically flat H-terminated Si(111) surfaces in aqueous solutions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Akihito Imanishi et al.
Langmuir : the ACS journal of surfaces and colloids, 23(26), 12966-12972 (2007-11-21)
The formation of self-assembled monolayers (SAMs) of adsorbed cationic or anionic surfactant molecules on atomically flat H-terminated Si(111) surfaces in aqueous solutions was investigated by in situ AFM measurements, using octyl trimethylammonium chloride (C8TAC), dodecyl trimethylammonium chloride (C12TAC), octadecyl trimethylammonium
S O Odeyinde et al.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS, 66(2), 215-223 (2012-10-13)
Interventional radiology has a principal role frequently replacing surgery in the multidisciplinary management of vascular malformations. Having noticed an initial high rate of complications, we examined our early experience to study in detail the outcomes and risks. We also present
Anne-Claude Crémieux et al.
The Journal of antimicrobial chemotherapy, 74(9), 2666-2675 (2019-07-03)
In a new experimental model of carbapenemase-producing Klebsiella pneumoniae osteomyelitis we evaluated the efficacy of colistin alone and in various combinations and examined the emergence of colistin-resistant strains and cross-resistance to host defence peptides (HDPs). KPC-99YC is a clinical strain
Amar Mukund et al.
Journal of vascular and interventional radiology : JVIR, 23(9), 1200-1206 (2012-07-27)
Balloon-occluded retrograde transvenous obliteration (BRTO) is a widely accepted treatment for gastric varices, but data are limited in regard to its role in the management of hepatic encephalopathy (HE). This study evaluated the efficacy of BRTO with foam sclerotherapy in
Pascal Chabrot et al.
Journal of endovascular therapy : an official journal of the International Society of Endovascular Specialists, 19(3), 439-449 (2012-07-14)
To investigate whether embolization with chitosan hydrogel (CH) with or without a sclerosant (sodium tetradecyl sulphate, STS) can induce chemical endothelial ablation and prevent endothelial recanalization in a rabbit model. Chitosan radiopaque thermogels were prepared using chitosan, β-glycerophosphate, iopamidol, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service