Skip to Content
Merck
All Photos(1)

Key Documents

291102

Sigma-Aldrich

Piperonylbutoxide

technical grade, 90%

Synonym(s):

2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
Beilstein:
288063
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

refractive index

n20/D 1.498 (lit.)

bp

155 °C/0.3 mmHg (lit.)

solubility

water: slightly soluble 0.0289 g/L at 20.4 °C

density

1.059 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOCCOCc1cc2OCOc2cc1CCC

InChI

1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3

InChI key

FIPWRIJSWJWJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Piperonylbutoxide is a safe insecticide for household and field. Effect of piperonyl butoxide, a synthetic methylenedioxyphenyl inhibitor of cytochrome(s) P450, on the toxicity of organophosphate insecticides to cladoceran test species (Ceriodaphnia dubia, Daphnia magna and Daphnia pulex) has been investigated.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piperonyl butoxide, a new and safe insecticide for the household and field.
W E DOVE
Pests and their control, 15(9), 30-30 (1947-09-01)
G T Ankley et al.
Ecotoxicology and environmental safety, 21(3), 266-274 (1991-06-01)
Experiments were conducted to determine the effects of piperonyl butoxide, a synthetic methylenedioxyphenyl inhibitor of cytochrome(s) P450, on the toxicity of organophosphate insecticides to three cladoceran test species: Ceriodaphnia dubia. Daphnia magna, and Daphnia pulex. Coadministration of piperonyl butoxide effectively
Graham D Moores et al.
Pest management science, 68(5), 795-800 (2012-01-10)
Previous work has characterised pyrethroid resistance in pollen beetle (Meligethes aeneus F.) as principally an oxidative mechanism. Piperonyl butoxide (PBO) can synergise this resistance in the field, but its effects on the honey bee are thought to be unacceptable. A
Petek Piner et al.
Environmental toxicology and pharmacology, 33(3), 414-420 (2012-03-06)
The aim of this study was to investigate the toxic effects of pyrethroid pesticide lambda-cyhalothrin in the presence of piperonyl butoxide as a modulator in the liver of juvenile Oreochromis niloticus. LC(50) (96h) value of lambda-cyhalothrin was determined as 2.901μg/L
Ranil Waliwitiya et al.
Journal of medical entomology, 49(3), 614-623 (2012-06-12)
The biochemical mechanisms underlying the increased toxicity of several plant essential oils (thymol, eugenol, pulegone, terpineol, and citronellal) against fourth instar of Aedes aegypti L. when exposed simultaneously with piperonyl butoxide (PBO) were examined. Whole body biotransformational enzyme activities including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service