All Photos(2)
About This Item
Linear Formula:
CH3(CH2)3COOC2H5
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744680
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
refractive index
n20/D 1.401 (lit.)
bp
144-145 °C (lit.)
density
0.875 g/mL at 25 °C (lit.)
SMILES string
CCCCC(=O)OCC
InChI
1S/C7H14O2/c1-3-5-6-7(8)9-4-2/h3-6H2,1-2H3
InChI key
ICMAFTSLXCXHRK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Ethyl valerate is also known as the green apple flavor. Microwave spectrum of ethyl valerate, observed by molecular beam Fourier transform microwave spectroscopy, has been reported.
Application
Ethyl valerate has wide applications in the areas of food, pharmaceuticals and cosmetics industries.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
93.2 °F - closed cup
Flash Point(C)
34 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jordan M Ross et al.
Bio-protocol, 8(18), e3013-e3013 (2018-09-20)
Classical fear conditioning typically involves pairing a discrete cue with a foot shock. Quantifying behavioral freezing to the learned cue is a crucial assay for neuroscience studies focused on learning and memory. Many paradigms utilize discrete stimuli such as tones;
H Mouhib et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(5), 1297-1301 (2012-03-02)
We report on the microwave spectrum of ethyl valerate, C(4)H(9)-COO-C(2)H(5), observed by molecular beam Fourier transform microwave spectroscopy (MB-FTMW). Highly accurate rotational and centrifugal distortion constants of the two most abundant conformers were determined. Their structure was investigated by comparison
Jordan M Ross et al.
Frontiers in behavioral neuroscience, 13, 164-164 (2019-08-06)
The olfactory bulb (OB) receives significant cholinergic innervation and widely expresses cholinergic receptors. While acetylcholine (ACh) is essential for olfactory learning, the exact mechanisms by which ACh modulates olfactory learning and whether it is specifically required in the OB remains
Synthesis of the 'green apple ester'ethyl valerate in organic solvents by Candida rugosa lipase immobilized in MBGs in organic solvents: Effects of immobilization and reaction parameters.
Raghavendra T, et al.
Journal of Molecular Catalysis. B, Enzymatic, 63(1), 31-38 (2010)
José Manuel Muñoz-Redondo et al.
Journal of agricultural and food chemistry, 65(13), 2768-2775 (2017-03-14)
A quantitative approach using HS-SPME-GC-MS was performed to investigate the ester changes related to the second fermentation in bottle. The contribution of the type of base wine to the final wine style is detailed. Furthermore, a discriminant model was developed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service