Skip to Content
Merck
All Photos(1)

Key Documents

288284

Sigma-Aldrich

Dicyclohexylphenylphosphine

95%

Synonym(s):

Phenyldicyclohexylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2PC6H5
CAS Number:
Molecular Weight:
274.38
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

60-61 °C (lit.)

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)c3ccccc3

InChI

1S/C18H27P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1,4-5,10-11,17-18H,2-3,6-9,12-15H2

InChI key

VPLLTGLLUHLIHA-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatyana I Rokitskaya et al.
Biochimica et biophysica acta. Biomembranes, 1863(1), 183483-183483 (2020-10-02)
To clarify the contribution of charge delocalization in a lipophilic ion to the efficacy of its permeation through a lipid membrane, we compared the behavior of alkyl derivatives of triphenylphosphonium, tricyclohexylphosphonium and trihexylphosphonium both in natural and artificial membranes. Exploring

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service