284297
2-Mercaptoimidazole
98%
Synonym(s):
2-Imidazolethiol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H4N2S
CAS Number:
Molecular Weight:
100.14
Beilstein:
105775
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
mp
228-231 °C (lit.)
solubility
methanol: soluble 50 mg/mL, clear to slightly hazy, colorless to yellow
SMILES string
Sc1ncc[nH]1
InChI
1S/C3H4N2S/c6-3-4-1-2-5-3/h1-2H,(H2,4,5,6)
InChI key
OXFSTTJBVAAALW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The kinetics of inhibition on mushroom tyrosinase by 2-mercaptoimidazole has been investigated. The pH dependent surface-enhanced Raman scattering (SERS) spectra of biologically and industrially significant, 2-mercaptoimidazole molecule at 1.0×10-10M concentration have been investigated.
Application
2-Mercaptoimidazole has been used in chemical modification of silica gel for selective separation of mercury(II) from an aqueous solution.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Subhendu Chandra et al.
Journal of colloid and interface science, 399, 33-45 (2013-04-02)
The pH dependent surface-enhanced Raman scattering (SERS) spectra of biologically and industrially significant, 2-mercaptoimidazole (2-MI) molecule at 1.0×10(-10)M concentration have been investigated. The pH dependent SERS spectra are compared with the corresponding normal Raman spectra (NRS) of the molecule. The
Lívia P L Matos et al.
Clinical and experimental pharmacology & physiology, 45(4), 326-334 (2017-11-08)
Na+ /I- symporter (NIS) transports iodide into thyrocytes, a fundamental step for thyroid hormone biosynthesis. Our aim was to evaluate NIS regulation in different status of goitrogenesis and its underlying mechanisms. Wistar rats were treated with methimazole (MMI) for 5
Chin-Feng Chan et al.
Bioorganic & medicinal chemistry, 22(9), 2809-2815 (2014-04-08)
In this study, we synthesized 4 methimazole (2-mercapto-1-methylimidazole, MMI) derivatives. The kinetics of inhibition on mushroom tyrosinase by methimazole and its derivatives were investigated. The results indicated that tert-butyl 3-methyl-2-sulfanylidene-2,3-dihydro-1H-imidazole-1-carboxylate (compound 3; 3), 2-mercaptoimidazole (MI; compound 1; 1) and MMI
2-Mercaptoimidazole covalently bonded to a silica gel surface for the selective separation of mercury (II) from an aqueous solution.
Filho NLD and Gushikem Y.
Separation Science and Technology, 32(15), 2535-2545 (1997)
C Kimblin et al.
Inorganic chemistry, 39(19), 4240-4243 (2001-02-24)
The bis(2-mercapto-1-methylimidazolyl)(pyrazolyl)hydroborato derivatives [pzBmMe]2Zn, [pzBmMe]2Co, and [pzBmMe]2Cd have been isolated and structurally characterized by X-ray diffraction. Despite their common [pzBmMe]2M composition, each of these complexes adopts a different structure. Thus, (i) the zinc complex exhibits a tetrahedral Zn[S4] structure in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service