279412
Sodium formate-13C
99 atom % 13C
Synonym(s):
Formic-13C acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
99 atom % 13C
form
solid
mp
259-262 °C (lit.)
mass shift
M+1
SMILES string
[Na+].[O-][13CH]=O
InChI
1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1/i1+1;
InChI key
HLBBKKJFGFRGMU-YTBWXGASSA-M
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nature genetics, 51(6), 990-998 (2019-05-28)
The histone acetyl reader bromodomain-containing protein 4 (BRD4) is an important regulator of chromatin structure and transcription, yet factors modulating its activity have remained elusive. Here we describe two complementary screens for genetic and physical interactors of BRD4, which converge
Science (New York, N.Y.), 368(6488), 283-290 (2020-04-18)
Metabolons, multiprotein complexes consisting of sequential enzymes of a metabolic pathway, are proposed to be biosynthetic "hotspots" within the cell. However, experimental demonstration of their presence and functions has remained challenging. We used metabolomics and in situ three-dimensional submicrometer chemical
Journal of animal science, 90 Suppl 4, 212-214 (2013-02-13)
Two studies, arranged according to a 4 × 4 Latin square design, were conducted to assess effects of dietary acidification on fungal 3-phytase (PHY) efficacy in growing pigs. In Exp. 1, effects of supplementing 500 units/kg feed of PHY and
Applied biochemistry and biotechnology, 169(7), 2038-2048 (2013-01-30)
The use of lignocellulosic materials as substrate for bioethanol production is considered a cost-effective approach to make the biofuel production process economically sustainable. However, lignocellulosic hydrolysis releases toxic compounds such as weak acids which inhibit microorganism growth and ethanol production.
Journal of bacteriology, 195(7), 1456-1462 (2013-01-22)
Hydrogenotrophic methanogenic Archaea are defined by an H2 requirement for growth. Despite this requirement, many hydrogenotrophs are also capable of growth with formate as an electron donor for methanogenesis. While certain responses of these organisms to hydrogen availability have been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service