Skip to Content
Merck
All Photos(1)

Key Documents

27395

Sigma-Aldrich

1,4-Cineole

technical, mixture of isomers, ≥85% (GC)

Synonym(s):

1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
104974
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥85% (GC)

form

liquid

composition

1,4-cineol, ~75%
1,8-cineol, ~15%

refractive index

n20/D 1.445 (lit.)
n20/D 1.447

bp

65 °C/16 mmHg (lit.)

mp

−46 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CC(C)C12CCC(C)(CC1)O2

InChI

1S/C10H18O/c1-8(2)10-6-4-9(3,11-10)5-7-10/h8H,4-7H2,1-3H3/t9-,10+

InChI key

RFFOTVCVTJUTAD-AOOOYVTPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Cineole is present in eucalyptus oil. It activated both human TRPA1 (a sensor of noxious cold) and human TRPM 8 (a thermosensitive receptor). It is a natural monoterpene that inhibits plant growth.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ji et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 22(8), 489-492 (1997-08-01)
The essential oils from the dried stems of Ephedra sinica, E. intermedia and E. equisetina were analyzed by GC-MS qualitatively and GC quantitatively. One hundred and twenty-seven constituents were identified, l-alpha-terpineol (31.64%) in E. sinica, 1,4-cineole (12.80%) in E. intermedia
W Liu et al.
Applied and environmental microbiology, 59(11), 3889-3893 (1993-11-01)
A cytochrome P-450-dependent monooxygenase system that catalyzes the stereospecific hydroxylation of the monoterpene substrate 1,4-cineole was demonstrated in cell-free preparations of Bacillus cereus UI-1477. 1,4-Cineole hydroxylations were catalyzed by a 100,000 x g (1-h)-centrifuging soluble, hexane-inducible enzyme that activated and
M Miyazawa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 31(10), 713-723 (2001-11-07)
1. Oxidation of 1,4-cineole, a monoterpene cyclic ether, was studied in rat and human liver microsomes and recombinant cytochrome P450 (P450 or CYP) enzymes expressed in insect cells in which human P450 and NADPH-P450 reductase cDNAs have been introduced. On
Patrícia Bezerra Gomes et al.
Pharmacology, biochemistry, and behavior, 96(3), 287-293 (2010-07-31)
Recent studies have shown that some monoterpenes exert anxiolytic- and depressant-like actions, however, these effects from monoterpene 1,4-cineole are still unknown. This work aimed to study the effects of 1,4-cineole in classic animal models for depression- and anxiety-like behavior, specifically
Abdul Ahad et al.
Current drug delivery, 8(2), 213-224 (2011-01-18)
The purpose of this study was to investigate the effectiveness and mechanism(s) of percutaneous absorption of propranolol hydrochloride (PHCL) across rat and human cadaver skin using seven novel terpenes with reference to marker terpene 1,8-cineole. In-vitro skin permeation studies were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service