Skip to Content
Merck
All Photos(1)

Key Documents

273643

Sigma-Aldrich

2-Octynal

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C≡CCHO
CAS Number:
Molecular Weight:
124.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

74-76 °C/15 mmHg (lit.)

density

0.871 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CCCCCC#CC=O

InChI

1S/C8H12O/c1-2-3-4-5-6-7-8-9/h8H,2-5H2,1H3

InChI key

DUUGWLGPIZCNLM-UHFFFAOYSA-N

Application

2-Octynal was used in the synthesis of (R)- and (S)-argentilactone via enantioselective catalytic allylation (ECA) and ring-closing metathesis pathways.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelo de Fatima et al.
Bioorganic & medicinal chemistry, 12(20), 5437-5442 (2004-09-25)
Concise total syntheses of (R)- and (S)-argentilactone have been developed via enantioselective catalytic allylation (ECA) and ring-closing metathesis pathways (four steps, 39% overall yield and 82-84% ee) from 2-octynal and their in vitro activity against cancer cells is described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service