Skip to Content
Merck
All Photos(1)

Key Documents

261696

Sigma-Aldrich

Methyl phenyl sulfoxide

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SOC6H5
CAS Number:
Molecular Weight:
140.20
Beilstein:
1904976
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

crystals

refractive index

n20/D 1.5775 (lit.)

bp

139-140 °C/14 mmHg (lit.)

mp

26-29 °C (lit.)

functional group

sulfoxide

storage temp.

2-8°C

SMILES string

CS(=O)c1ccccc1

InChI

1S/C7H8OS/c1-9(8)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

JXTGICXCHWMCPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-methyl phenyl sulfoxide is an important pharmaceutical intermediate.

Application

Methyl phenyl sulfoxide has been used in the synthesis of:
  • isotopically labelled 1, 3-dithiane, a useful labeled synthon
  • nelfinavir, potent HIV-protease inhibitor
  • sulfoximines and in studies on solvent-water partitioning

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M H Abraham et al.
Journal of pharmaceutical sciences, 83(8), 1085-1100 (1994-08-01)
A general linear solvation energy equation has been used to analyze published partition coefficients in the systems water-octanol (613 solutes), water-hexadecane (370 solutes), water-alkane (200 solutes), and water-cyclohexane (170 solutes). The descriptors used in the equation are R2, an excess
Sadagopan Raghavan et al.
The Journal of organic chemistry, 75(2), 498-501 (2009-12-17)
An asymmetric synthesis of nelfinavir is described starting from acrolein and (S)-methyl phenyl sulfoxide. The key features include (a) stereoselective preparation of a beta-protected amino-gamma,delta-unsaturated sulfoxide by the reaction of an alpha-sulfinyl carbanion with an unsaturated t-butyl sulfinylimine, (b) stereoselective
Nathalie Sauvonnet et al.
The Journal of cell biology, 168(1), 155-163 (2004-12-30)
Endocytosis is critical for many cellular functions. We show that endocytosis of the common gammac cytokine receptor is clathrin independent by using a dominant-negative mutant of Eps15 or RNA interference to knock down clathrin heavy chain. This pathway is synaptojanin
Aldrichimica Acta, 18, 3-3 (1985)
Rodolfo A Martinez et al.
Journal of labelled compounds & radiopharmaceuticals, 57(5), 338-341 (2014-05-28)
The 1,3-dithiane is a protected formaldehyde anion equivalent that could serve as a useful labeled synthon. We report a facile synthesis of 1,3-[2-(13)C]- and 1,3-[2-(13)C, 2-(2)H2]dithiane in two steps from [(13)C]- or [(13) C, (2)H3 ]methyl phenyl sulfoxide. We have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service