Skip to Content
Merck
All Photos(1)

Key Documents

257591

Sigma-Aldrich

4,4′-(Hexafluoroisopropylidene)diphenol

97%

Synonym(s):

2,2-Bis(4-hydroxyphenyl)hexafluoropropane, Bisphenol AF, Hexafluorobisphenol A

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C(C6H4OH)2
CAS Number:
Molecular Weight:
336.23
Beilstein:
1891568
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

160-163 °C (lit.)

SMILES string

Oc1ccc(cc1)C(c2ccc(O)cc2)(C(F)(F)F)C(F)(F)F

InChI

1S/C15H10F6O2/c16-14(17,18)13(15(19,20)21,9-1-5-11(22)6-2-9)10-3-7-12(23)8-4-10/h1-8,22-23H

InChI key

ZFVMWEVVKGLCIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Zhang et al.
Toxicology letters, 283, 32-38 (2017-11-13)
The binding of bisphenol A (BPA) and its halogenated derivatives (halogenated BPAs) to mouse peroxisome proliferator-activated receptor α ligand binding domain (mPPARα-LBD) was examined by a combination of in vitro investigation and in silico simulation. Fluorescence polarization (FP) assay showed
John Moreman et al.
Environmental science & technology, 51(21), 12796-12805 (2017-10-11)
Bisphenol A (BPA), a chemical incorporated into plastics and resins, has estrogenic activity and is associated with adverse health effects in humans and wildlife. Similarly structured BPA analogues are widely used but far less is known about their potential toxicity
Tengfei Wang et al.
Talanta, 178, 481-490 (2017-11-16)
Using sulfonated and fluorinated poly (arylene ether ketone) comprising functional strong coordination group benzimidazole (SPAEK-F-BI) as a template film, a novel fabrication method of cobalt nanoflowers (CoNFs) and non-enzymatic glucose electrochemical sensor was developed in this work. After the precursors
Lalith Perera et al.
Toxicology in vitro : an international journal published in association with BIBRA, 44, 287-302 (2017-07-29)
Bisphenol A (BPA), bisphenol AF (BPAF), and bisphenol S (BPS) are well known endocrine disruptors. Previous in vitro studies showed that these compounds antagonize androgen receptor (AR) transcriptional activity; however, the mechanisms of action are unclear. In the present study
Hiroyuki Okazaki et al.
Biological & pharmaceutical bulletin, 40(11), 1909-1916 (2017-11-03)
Bisphenols are endocrine disruptors that are widely found in the environment. Accumulating experimental evidence suggests an adverse interaction between bisphenols and estrogen signaling. Most studies have performed experiments that focused on estrogen receptor (ER) engagement by bisphenols. Therefore, the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service