Skip to Content
Merck
All Photos(3)

Key Documents

255920

Sigma-Aldrich

1,1′-(Azodicarbonyl)dipiperidine

99%

Synonym(s):

1,1′-Azobis(N,N-pentamethyleneformamide), ADD, Azodicarboxylic acid dipiperidide, NSC 356027, SR 4077

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N4O2
CAS Number:
Molecular Weight:
252.31
Beilstein:
261917
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

134-136 °C (lit.)

functional group

azo

SMILES string

O=C(\N=N\C(=O)N1CCCCC1)N2CCCCC2

InChI

1S/C12H20N4O2/c17-11(15-7-3-1-4-8-15)13-14-12(18)16-9-5-2-6-10-16/h1-10H2/b14-13+

InChI key

OQJBFFCUFALWQL-BUHFOSPRSA-N

Application

Reactant for preparation of:
  • Polyfluoroalkylated tripyrazolylmethane ligands

  • (-)-Hygromycin A via Mitsunobu glycosylation
  • Optically active α,α-disubstituted amino acids via Mitsunobu reaction
  • Aza-β-lactams through [2+2] cycloaddition reactions
  • Glycosyl disulfides
  • Pyridine ether PPAR agonists
  • S-glycosyl amino acid building blocks for combinatorial neoglycopeptide synthesis
  • Histamine H3 receptor antagonists

Reactant for:
  • Mitsunobu inversion reactions
Used in a study of the copper-catalyzed addition of aryboronic acids to azodicarboxyl derivatives providing aryl-substituted hydrazides.
Widely used reagent for the Mitsunobu reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dougherty, J.M.; et al.
Tetrahedron, 61, 6218-6218 (2005)
Tetrahedron Letters, 34, 1639-1639 (1993)
Takeshi Uemura et al.
The Journal of organic chemistry, 70(21), 8631-8634 (2005-10-08)
The addition of arylboronic acids 1 to azodicarboxylates 2 in the presence of a catalytic amount of a copper salt under mild reaction conditions gives aryl-substituted hydrazines 3 in high yields. The reaction is tolerant of a wide variety of
M A Baker et al.
Radiation research, 113(2), 346-355 (1988-02-01)
The mechanism of radiosensitization by diazenedicarboxylic acid bis(N),N-piperidide (SR 4077), a less toxic analog of diamide, was studied using Chinese hamster ovary cells. SR 4077 gave an average SER of 1.58 for postirradiation incubations of 0.5, 1.0, or 2.0 h.
E A Bump et al.
International journal of radiation oncology, biology, physics, 29(2), 249-253 (1994-05-15)
To determine whether biological effects of radiation, such as apoptosis, that differ from classical clonogenic cell killing, can be modified with agents that would not be expected to modify classical clonogenic cell killing. This would expand the range of potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service