Skip to Content
Merck
All Photos(3)

Key Documents

255718

Sigma-Aldrich

Cesium fluoride

99.99% trace metals basis

Synonym(s):

NSC 84270

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsF
CAS Number:
Molecular Weight:
151.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
solid
solubility:
methanol: slightly soluble(lit.)
pyridine: insoluble(lit.)
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

solid

impurities

≤150.0 ppm Trace Metal Analysis

mp

682 °C (lit.)

solubility

methanol: slightly soluble(lit.)
pyridine: insoluble(lit.)

density

4.115 g/mL at 25 °C (lit.)

SMILES string

[F-].[Cs+]

InChI

1S/Cs.FH/h;1H/q+1;/p-1

InChI key

XJHCXCQVJFPJIK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of building blocks for synthesis of fluoroallylic compounds
  • Synthesis of alcohols via hydrolysis of alkyl silyl ethers at neutral pH in buffered mixed organic-aqueous solutions
  • Nucleophilic fluorination of alkynyliodonium salts to form fluorovinylic compounds
  • Nucleophilic aromatic substitution (SNAr) reactions
Used as a base in the Suzuki carbonylative coupling of enol triflates with vinyl boronic acids.[1]
Used in the successful synthesis of a single-crystal Dion-Jacobson phase, CsLaTa2O7, that has applications in photocatalysis and superconductivity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Adrenal gland

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Organic Chemistry, 2152-2152 (2007)
J Kypr et al.
International journal of biological macromolecules, 13(1), 9-13 (1991-02-01)
Ten DNA fragments containing self-complementary alternating sequences of adenine and thymine differing in length and the starting nucleotide were studied by c.d. spectroscopy. It was found that d(TATATATA) but not d(ATATATAT), d(TATATA), d(CTATATAG) or (dT-dA)20 isomerized into the unusual X-DNA
T C Melville et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(6), 1171-1176 (2001-06-23)
A procedure for directly fitting the potential energy curve of a diatomic molecule has been applied to the X1sigma+ ground states of LiCl, TlCl, RbF and CsF. Extensive, high-precision infrared and pure-rotational data sets for all isotopomers of the aforementioned
E D Risby et al.
Archives of general psychiatry, 48(6), 513-524 (1991-06-01)
As part of a study of the effects of lithium carbonate on neurochemical function in man, platelet and lymphocyte adenylate cyclase activity and lymphocyte beta-adrenergic receptor binding characteristics were determined before and after 2 weeks of lithium treatment in 10
E A Lesnik et al.
Journal of biomolecular structure & dynamics, 5(3), 601-614 (1987-12-01)
We have determined the 1H----3H exchange rate constants between water and C8H groups of purinic residues of alternating polynucleotides poly(dA-dT).poly(dA-dT), poly(dG-dC).poly(dG-dC) and poly(dA-dC).poly(dG-dT) as well as homopolynucleotides poly(dA).poly(dT) and poly(dG).poly(dC) in aqueous solutions with high-salt concentrations (3 M NaCl and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service