Skip to Content
Merck
All Photos(3)

Key Documents

255246

Sigma-Aldrich

Cresyl Violet perchlorate

Synonym(s):

Oxazine 9 perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O · HClO4
CAS Number:
Molecular Weight:
361.74
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

>96.5% (HPLC)

form

powder

λmax

602 nm

ε (extinction coefficient)

≥27000 at 267-273 nm
≥50000 at 599-605 nm
≥7000 at 316-322 nm

fluorescence

λem 602 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OCl(=O)(=O)=O.Nc1cc2OC3=CC(=N)C=CC3=Nc2c4ccccc14

InChI

1S/C16H11N3O.ClHO4/c17-9-5-6-13-14(7-9)20-15-8-12(18)10-3-1-2-4-11(10)16(15)19-13;2-1(3,4)5/h1-8,17H,18H2;(H,2,3,4,5)

InChI key

YZUJISGZMSLVAU-UHFFFAOYSA-N

General description

Cresyl Violet perchlorate, also known as oxazine 9, is a laser dye. Cresyl violet is a small diaminobenzooxazine with a moderately sized planar conjugated system.

Application

Among its many applications, Cresyl Violet can be used to stain for Nissl substance and nuclei in nervous tissue and the identification of Helicobacter. Cresyl Violet perchlorate has been used to make cresyl violet perchlorate-polymer composite thin films. It also has been used as a fluorescence standard.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

F. W. D. Rost
Fluorescence Microscopy, 2 (1992)
Estelle Rousselet et al.
Journal of visualized experiments : JoVE, (69)(69), doi:10-doi:10 (2012-11-22)
Stroke is the third cause of mortality and the leading cause of disability in the World. Ischemic stroke accounts for approximately 80% of all strokes. However, the thrombolytic tissue plasminogen activator (tPA) is the only treatment of acute ischemic stroke
Sung In Cho et al.
Bioelectromagnetics, 33(7), 568-574 (2012-04-13)
Our previous study has shown that an extremely low-frequency magnetic field (ELF-MF) induces nitric oxide (NO) synthesis by Ca(2+) -dependent NO synthase (NOS) in rat brain. The present study was designed to confirm that ELF-MF affects neuronal NOS (nNOS) in
Priscilla Hirst et al.
Neural plasticity, 2012, 197264-197264 (2012-07-14)
Recent studies suggest that exposure to only one component of audiovisual events can lead to cross-modal cortical activation. However, it is not certain whether such crossmodal recruitment can occur in the absence of explicit conditioning, semantic factors, or long-term associations.
Simon C Spanswick et al.
PloS one, 7(8), e43698-e43698 (2012-08-29)
Recent evidence suggests that the medial prefrontal cortex (MFC) is important for processing contextual information. Here we evaluate the performance of mice with MFC damage in a discrimination task that requires an association between an object and the context in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service