252557
4-Hydroxy-2-butanone
95%
Synonym(s):
2-Hydroxyethyl methyl ketone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HOCH2CH2COCH3
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
refractive index
n20/D 1.430 (lit.)
bp
73-76 °C/12 mmHg (lit.)
density
1.023 g/mL at 25 °C (lit.)
functional group
hydroxyl
ketone
SMILES string
CC(=O)CCO
InChI
1S/C4H8O2/c1-4(6)2-3-5/h5H,2-3H2,1H3
InChI key
LVSQXDHWDCMMRJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Hydroxy-2-butanone is a beta-hydroxy ketone used as a starting material to synthesize 1,3-butanediol.
4-Hydroxy-2-butanone is an important pharmaceutical intermediate. Gas phase reaction of the OH radicals with 4-hydroxy-2-butanone has been investigated by absolute rate method.
4-Hydroxy-2-butanone is an important pharmaceutical intermediate. Gas phase reaction of the OH radicals with 4-hydroxy-2-butanone has been investigated by absolute rate method.
Application
4-Hydroxy-2-butanone has been used in the synthesis of:
- 3-buten-2-one via dehydration over anatase-TiO2 catalyst
- 4-methyl-5-hydroxymethylthiazole
- (±)-lineatin
- mevalonic acid lactone
- verrucarin
- β-hydroxylactones
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
91.4 °F - closed cup
Flash Point(C)
33 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Recent advances in fermentative production of C4 diols and their chemo-catalytic upgrading to high-value chemicals
Varma AR, et al.
Chinese Journal of Catalysis, 52, 99-126 (2023)
Synthesis of 3-buten-2-one from 4-hydroxy-2-butanone over anatase-TiO2 catalyst.
Ichikawa N, et al.
Catalysis Communications, 6(1), 19-22 (2005)
The Journal of Organic Chemistry, 47, 929-929 (1982)
Journal of the American Chemical Society, 79, 2316-2316 (1957)
Studies on the biosynthesis of cholesterol. 5. Biosynthesis of squalene from DL-3-hydroxy-3-methyl [2-14C] pentano-5-lactone.
J W CORNFORTH et al.
The Biochemical journal, 69(1), 146-155 (1958-05-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service