Skip to Content
Merck
All Photos(3)

Key Documents

248835

Sigma-Aldrich

p-Toluenesulfonyl cyanide

technical grade, 95%

Synonym(s):

p-Tolylsulfonyl cyanide, Tosyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2CN
CAS Number:
Molecular Weight:
181.21
Beilstein:
2048159
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

95%

form

solid

bp

105-106 °C/1 mmHg (lit.)

mp

47-50 °C (lit.)

functional group

sulfone
thiocyanate

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)C#N

InChI

1S/C8H7NO2S/c1-7-2-4-8(5-3-7)12(10,11)6-9/h2-5H,1H3

InChI key

JONIMGVUGJVFQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Toluenesulfonyl cyanide is an electron-deficient nitrile. It undergoes hetero-Diels-Alder reaction with silyl enol ether of cyclohexenone to yield hydrolytically sensitive [4+2] adduct. It also undergoes facile [2+3] cycloaddition reaction with azides to form 5-sulfonyl tetrazoles. p-Toluenesulfonyl cyanide also reacts with alcohols in the presence of 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU) or 1,4-diazabicyclo[2.2.2]octane (DABCO) to yield sulfinates.

Application

p-Toluenesulfonyl cyanide can be used in:
  • The preparation of polyfunctional nitriles.
  • Free-radical cyanation of B-alkylcatecholboranes.
  • The synthesis of 4-hyroxypyridines by hetero-Diels-Alder reaction with 1,3-bis-silyl enol ethers.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A click chemistry approach to tetrazoles by Huisgen 1,3-dipolar cycloaddition: synthesis of 5-acyltetrazoles from azides and acyl cyanides.
Zachary P Demko et al.
Angewandte Chemie (International ed. in English), 41(12), 2113-2116 (2002-06-17)
Synthesis of 2-(Arylsulfonyl)-4-hydroxypyridines by Hetero-Diels-Alder Reaction of 1,3-Bis-Silyl Enol Ethers with Arylsulfonyl Cyanides
Thomas Emmrich, et al.
Synthesis, 2551-2555 (2006)
O-Sulfinylation of alcohols with methanesulfonyl cyanide or p-toluenesulfonyl cyanide.
Barton DHR, et al.
Tetrahedron, 47(44), 9167-9178 (1991)
Preparation of polyfunctional nitriles by the cyanation of functionalized organozinc halides with p-toluenesulfonyl cyanide.
Klement I, et al.
Tetrahedron Letters, 34(29), 4623-4626 (1993)
Radical-mediated alkenylation, alkynylation, methanimination, and cyanation of B-alkylcatecholboranes.
Arnaud-Pierre Schaffner et al.
Angewandte Chemie (International ed. in English), 45(35), 5847-5849 (2006-08-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service