Skip to Content
Merck
All Photos(3)

Key Documents

245615

Sigma-Aldrich

2-Bromo-4′-nitroacetophenone

95%

Synonym(s):

ω-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH2Br
CAS Number:
Molecular Weight:
244.04
Beilstein:
393567
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

94-99 °C (lit.)

functional group

bromo
ketone
nitro

SMILES string

[O-][N+](=O)c1ccc(cc1)C(=O)CBr

InChI

1S/C8H6BrNO3/c9-5-8(11)6-1-3-7(4-2-6)10(12)13/h1-4H,5H2

InChI key

MBUPVGIGAMCMBT-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Bromo-4′-nitroacetophenone was used to study the pKa of the histidine-34 imidazole.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R R Annand et al.
Biochemistry, 35(14), 4591-4601 (1996-04-09)
In bee venom phospholipase A2, histidine-34 probably functions as a Brønsted base to deprotonate the attacking water. Aspartate-64 and tyrosine-87 form a hydrogen bonding network with histidine-34. We have prepared mutants at these positions and studied their kinetic properties. The
A Parkinson et al.
The Journal of biological chemistry, 261(25), 11478-11486 (1986-09-05)
The alkylating agent 2-bromo-4'-nitroacetophenone (BrNAP) binds covalently to each of 10 isozymes of purified rat liver microsomal cytochrome P-450 (P-450a-P-450j) but substantially inhibits the catalytic activity of only cytochrome P-450c. Regardless of pH, incubation time, presence of detergents, or concentration
Dana Ortansa Dorohoi et al.
Molecules (Basel, Switzerland), 26(13) (2021-07-03)
Two 4-tolyl-1,2,4-triazol-1-ium methylids, namely 4-tolyl-1,2,4-triazol-1-ium-phenacylid and 4-tolyl-1,2,4-triazol-1-ium-4'-nitro-phenacylid, are studied from solvatochromic point of view in binary solvent mixtures of water with ethanol and water with methanol. The contributions (expressed in percent) of the universal and specific interactions are separated from
B Z Yu et al.
Biochemistry, 36(13), 3870-3881 (1997-04-01)
Interfacial catalytic constants for bee venom phospholipase A2 (bvPLA2) have been obtained for its action on vesicles of the anionic phospholipid 1,2-dimyristoylphosphatidylmethanol (DMPM) in the highly processive scooting mode. Spectroscopic measurements which directly measure transbilayer movement of membrane components show
Hao Wang et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(11), 1297-1302 (2018-07-31)
A new highly selective and visible colorimetric fluorescent probe (probe 1) was developed to detect hydrazine (N2H4) concentration in real water samples. As different concentrations of N2H4 were added, the color of the probe solution was graded gradually from colorless

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service