Skip to Content
Merck
All Photos(1)

Key Documents

243639

Sigma-Aldrich

Tungsten(IV) sulfide

powder, 2 μm, 99%

Synonym(s):

Tungsten disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WS2
CAS Number:
Molecular Weight:
247.97
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

particle size

2 μm

density

7.5 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

S=[W]=S

InChI

1S/2S.W

InChI key

ITRNXVSDJBHYNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tungsten(IV) sulfide (WS2) is a transition metal dichalcogenide with good catalytic, optical and electronic properties. WS2 can be used as a solid lubricant for a variety of applications such as aerospace, cutting tools and automotive systems.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Codeposition of inorganic fullerene-like WS2 nanoparticles in an electrodeposited nickel matrix under the influence of ultrasonic agitation
Garcia-Lecina E, et al.
Electrochimica Acta, 114, 859-867 (2013)
Performance of femtosecond laser-textured cutting tools deposited with WS2 solid lubricant coatings
Deng J, et al.
Surface and Coatings Technology, 222(1), 135-143 (2013)
Antispalling effect of WS2 nanoparticles on the lubrication of automotive gearboxes
Aldana PU, et al.
Tribology Transactions, 59(1), 178-188 (2016)
Maria O Zhukova et al.
Nanoscale research letters, 14(1), 225-225 (2019-07-11)
Time-resolved terahertz spectroscopy has become a common method both for fundamental and applied studies focused on improving the quality of human life. However, the issue of finding materials applicable in these systems is still relevant. One of the appropriate solution
WC/DLC/WS 2 nanocomposite coatings for aerospace tribology
Voevodin AA, et al.
Tribology Letters, 6(2), 75-78 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service