Skip to Content
Merck
All Photos(1)

Key Documents

241970

Sigma-Aldrich

Tetrapentylammonium bromide

≥99%

Synonym(s):

Tetra-n-pentylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4]4N(Br)
CAS Number:
Molecular Weight:
378.47
Beilstein:
3658171
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

mp

100-101 °C (lit.)

SMILES string

[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

InChI

1S/C20H44N.BrH/c1-5-9-13-17-21(18-14-10-6-2,19-15-11-7-3)20-16-12-8-4;/h5-20H2,1-4H3;1H/q+1;/p-1

InChI key

SPALIFXDWQTXKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tetrapentylammonium bromide is a quaternary ammonium salt with pentyl chains and a bromide counterion, which is generally used as a phase transfer catalyst.

Application

Tetrapentylammonium bromide can be used:
  • As a versatile structure-directing agent for the synthesis of Zeolite-like heterobimetallic cyanide frameworks.
  • As an alkylating agent for rhodium(I)-catalyzed alkylation reaction of benzylic amines and for N-alkylation of azaheterocycles.
  • As a precursor to prepare a Cobalt-complex, {[(Pentyl)4N]3CoBr3}Cl2, which is used as a catalyst in the synthesis of multiwalled carbon nanotubes (MWCNTs).


Electrochemical synthesis of stable graphite intercalation compounds (GICs) containing tetrapentylammonium cations has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel catalyst based on Co-complex to prepare MWCNT.
Alonso-Nu?ez G, et al.
Materials Letters, 109, 163-166 (2013)
Quaternary ammonium salts as highly efficient and green alkylating agents for N-alkylation of azaheterocycles under microwave irradiation.
Khalafi-Nezhad A, et al.
Journal of the Iranian Chemical Society, 5(1), S40-S46 (2008)
Quaternary Ammonium Salts as Alkylating Reagents in C-H Activation Chemistry.
Spettel M, et al.
Organic Letters, 19(16), 4287-4290 (2017)
Chemical modification of polymers via phase transfer catalysis
Nishikubo T.
Handbook of Phase Transfer Catalysis, 480-509 (1997)
Simple Quaternary Ammonium Ions R 4 N+(R= nPr, nBu, nPen) as Versatile Structure Directors for the Synthesis of Zeolite-Like, Heterobimetallic Cyanide Frameworks.
Poll EM, et al.
Journal of Solid State Chemistry, 152(1), 286-301 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service