Skip to Content
Merck
All Photos(1)

Documents

241806

Sigma-Aldrich

(R)-(−)-2,2-Dimethyl-1,3-dioxolane-4-methanol

98%

Synonym(s):

2,3-Isopropylidene-sn-glycerol, L-α,β-Isopropylideneglycerol, L-2,3-O-isopropylidene-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Beilstein:
80117
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −13.7°, neat

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.434 (lit.)

bp

72-73 °C/8 mmHg (lit.)

density

1.062 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)OC[C@@H](CO)O1

InChI

1S/C6H12O3/c1-6(2)8-4-5(3-7)9-6/h5,7H,3-4H2,1-2H3/t5-/m1/s1

InChI key

RNVYQYLELCKWAN-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol may be used to prepare:
  • (R)-2,2-Dimethyl-4-benzoxymethyl-1,3-dioxolane.
  • ((S)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulfonate)-1,3-dioxolane.
  • (S)-Glyceraldehyde acetonide.
  • Tetraoxaspiroundecanes, diglycerides, glyceryl phosphates, and spiropyrans.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S-Aryl-L-cysteine S,S-dioxides: design, synthesis, and evaluation of a new class of inhibitors of kynureninase
Rajesh K. Dua,
Journal of the American Chemical Society, 115, 4, 1264-1270 (1993)
European Journal of Organic Chemistry, 4805-4805 (2006)
René Pemha et al.
Marine drugs, 18(2) (2020-02-23)
A series of novel substituted 1-O-alkylglycerols (AKGs) containing methoxy (8), gem-difluoro (9), azide (10) and hydroxy (11) group at 12 position in the alkyl chain were synthesized from commercially available ricinoleic acid (12). The structures of these new synthesized AKGs
Synthesis of a `reverse ester? analogue of 1,2-sn-diglycerides from (S)-1,2-di-O-isoprophylideneglycerol; efficient, stereospecific nucleophilic displacement via a triflate at glycerol C-2
Bernard T Golding, et al.
Tetrahedron Letters, 34, 6459-6462 (1993)
Enantioselective synthesis of N1999A2.
Ji N, et al.
Journal of the American Chemical Society, 128(46), 14825-14825 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service