Skip to Content
Merck
All Photos(1)

Key Documents

240656

Sigma-Aldrich

1,2-Dibromoethane

≥99%

Synonym(s):

EDB, Ethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2Br
CAS Number:
Molecular Weight:
187.86
Beilstein:
605266
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~6.5 (vs air)

vapor pressure

11.7 mmHg ( 25 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

131-132 °C (lit.)

mp

8-11 °C (lit.)

solubility

water: soluble 250 part
alcohol: miscible(lit.)
diethyl ether: miscible(lit.)

density

2.18 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrCCBr

InChI

1S/C2H4Br2/c3-1-2-4/h1-2H2

InChI key

PAAZPARNPHGIKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dibromoethane (DBE) is an organobromine compound that is used as an ‘entrainment reagent′ to activate magnesium in Grignard reagents. It is also a useful reagent for activating zinc. DBE is used as an intermediate in the synthesis of dyes, pesticides and pharmaceuticals.

Application

1,2-Dibromoethane can be used as a reagent to prepare:
  • Vinyl bromide and 1,2-disubstituted ethane derivatives.
  • Vinyl aromatic compounds by palladium-catalyzed cross-coupling reaction with various arylboronic acids via in situ formation of vinyl bromide.
  • 2-arylsulfonyl hydrazones by reacting with aryldiazonium tetrafluoroborates, sulfur dioxide and hydrazines.

It can also be used as a bromine source in organic synthesis to brominate carbanions.

Other Notes

May darken in storage

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 2-Dibromoethane
Maynard GD
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
1, 2-dibromoethane-A versatile reagent in organic synthesis
Aluri BR
Synlett, 2008(10), 1579-1580 (2008)
Maya G Deshmukh et al.
Structure (London, England : 1993), 29(8), 823-833 (2021-06-24)
There is a clinical need for direct-acting antivirals targeting SARS-CoV-2, the coronavirus responsible for the COVID-19 pandemic, to complement current therapeutic strategies. The main protease (Mpro) is an attractive target for antiviral therapy. However, the vast majority of protease inhibitors
Saturation vapor pressures and transition enthalpies of low-volatility organic molecules of atmospheric relevance: from dicarboxylic acids to complex mixtures.
Merete Bilde et al.
Chemical reviews, 115(10), 4115-4156 (2015-05-02)
A four-component reaction of aryldiazonium tetrafluoroborates, sulfur dioxide, 1,2-dibromoethane, and hydrazines
Zheng D, et al.
Organic & Biomolecular Chemistry, 13(41), 10370-10375 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service