Skip to Content
Merck
All Photos(1)

Key Documents

24045

Sigma-Aldrich

2-(4-Chlorobutyl)-1,3-dioxolane

≥97.0% (GC)

Synonym(s):

5-Chloro-n-valeraldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13ClO2
CAS Number:
Molecular Weight:
164.63
Beilstein:
4304948
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.457

bp

56-58 °C/0.1 mmHg (lit.)

density

1.109 g/mL at 20 °C (lit.)

functional group

chloro
ether

SMILES string

ClCCCCC1OCCO1

InChI

1S/C7H13ClO2/c8-4-2-1-3-7-9-5-6-10-7/h7H,1-6H2

InChI key

GFTDGDAKGDUYGO-UHFFFAOYSA-N

Related Categories

Application

2-(4-Chlorobutyl)-1,3-dioxolane was used in the synthesis of C-chlorobutylpyrogallol[4]arene.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott J Dalgarno et al.
Chemical communications (Cambridge, England), (36)(36), 3803-3805 (2006-09-14)
Two lower rim halogenated pyrogallol[4]arenes have been synthesised and structurally characterised as either bi-layer or hexameric nano-capsule motifs depending upon chain length and functionalisation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service