Skip to Content
Merck
All Photos(2)

Key Documents

236306

Sigma-Aldrich

Allyl chloride

ReagentPlus®, 99%

Synonym(s):

3-Chloro-1-propene, Chlorallylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2Cl
CAS Number:
Molecular Weight:
76.52
Beilstein:
635704
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.6 (vs air)

Quality Level

vapor pressure

20.58 psi ( 55 °C)
5.71 psi ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

expl. lim.

11.2 %

refractive index

n20/D 1.414 (lit.)

bp

44-46 °C (lit.)

mp

−130 °C (lit.)

solubility

alcohol: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
petroleum ether: miscible(lit.)
water: slightly soluble(lit.)

density

0.939 g/mL at 25 °C (lit.)

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

ClCC=C

InChI

1S/C3H5Cl/c1-2-3-4/h2H,1,3H2

InChI key

OSDWBNJEKMUWAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetcis of epoxidation of allyl chloride with hydrogen peroxide has been reported.

Application

Allyl chloride has been employed:
  • as precursor for generation of allyl radicals by laser flash photolysis at 193nm
  • in the synthesis of α,ω-difunctional chemical intermediates via the cross-metathesis reactions

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-25.6 °F

Flash Point(C)

-32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars Seidel et al.
Molecules (Basel, Switzerland), 18(11), 13608-13622 (2013-11-07)
Photochemically driven reactions involving unsaturated radicals produce a thick global layer of organic haze on Titan, Saturn's largest moon. The allyl radical self-reaction is an example for this type of chemistry and was examined at room temperature from an experimental
Epoxidation of allyl chloride with hydrogen peroxide catalyzed by titanium silicalite 1.
Gao H, et al.
Applied Catalysis A: General, 138(1), 27-38 (1996)
Cross-metathesis reactions of allyl chloride with fatty acid methyl esters: Efficient synthesis of a, ?-difunctional chemical intermediates from renewable raw materials.
Jacobs T, et al.
Applied Catalysis A: General, 353(1), 32-35 (2009)
Shin-ichi Ikeda et al.
Chemical communications (Cambridge, England), (4)(4), 457-459 (2006-02-24)
The Ni-catalyzed coupling of allyl chlorides and enynes has been developed; the cyclization of enynes was triggered by the addition of pi-allylnickel species to the alkyne part, followed by the incorporation of the alkene part.
Justin M Chalker et al.
Chemical communications (Cambridge, England), (25)(25), 3714-3716 (2009-06-27)
Multiple, complementary methods are reported for the chemical conversion of cysteine to S-allyl cysteine on protein surfaces, a useful transformation for the exploration of olefin metathesis on proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service