Skip to Content
Merck
All Photos(1)

Key Documents

235210

Sigma-Aldrich

(1R,2S)-(−)-N-Methylephedrine

99%

Synonym(s):

(−)-(1R,2S)-2-Dimethylamino-1-phenylpropanol, (−)-N-Methylephedrine, [R-(R*,S*)]-α-[1-(Dimethylamino)ethyl]benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(CH3)N(CH3)2
CAS Number:
Molecular Weight:
179.26
Beilstein:
3031197
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]21/D −29.2°, c = 5 in methanol

mp

86-88 °C (lit.)

functional group

amine
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

C[C@@H]([C@H](O)c1ccccc1)N(C)C

InChI

1S/C11H17NO/c1-9(12(2)3)11(13)10-7-5-4-6-8-10/h4-9,11,13H,1-3H3/t9-,11-/m0/s1

InChI key

FMCGSUUBYTWNDP-ONGXEEELSA-N

Application

Resolving agent. Precursor to chiral supporting electrolytes, phase-transfer catalysts, and reducing agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry Letters (Jpn), 981-981 (1980)
Angewandte Chemie (International Edition in English), 23, 165-165 (1984)
Guoyi Ma et al.
The Journal of pharmacology and experimental therapeutics, 322(1), 214-221 (2007-04-05)
Ephedra species of plants have both beneficial and adverse effects primarily associated with the presence of ephedrine alkaloids. Few reports have appeared that examine the direct actions of ephedrine alkaloids on human subtypes of adrenergic receptors (ARs). In the present
María José Ojeda-Montes et al.
Future medicinal chemistry, 9(18), 2129-2146 (2017-11-28)
Extracts from Ephedra species have been reported to be effective as antidiabetics. A previous in silico study predicted that ephedrine and five ephedrine derivatives could contribute to the described antidiabetic effect of Ephedra extracts by inhibiting dipeptidyl peptidase IV (DPP-IV).
H J Lee et al.
Journal of obstetrics and gynaecology : the journal of the Institute of Obstetrics and Gynaecology, 30(6), 563-566 (2010-08-13)
No information is currently available on the safety of methylephedrine, a component of various cold medications available in South Korea. With previous approval by an Institutional Review Board, 349 women inadvertently exposed to methylephedrine during the 1st trimester of pregnancy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service