Skip to Content
Merck
All Photos(1)

Key Documents

229474

Sigma-Aldrich

Bismuth(III) iodide

≥99.998% trace metals basis

Synonym(s):

Bismuth triiodide, Triiodobismuthane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiI3
CAS Number:
Molecular Weight:
589.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

Quality Level

Assay

≥99.998% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: bismuth

impurities

≤15.0 ppm Trace Metal Analysis

mp

408 °C (lit.)

density

5.78 g/mL at 25 °C (lit.)

SMILES string

I[Bi](I)I

InChI

1S/Bi.3HI/h;3*1H/q+3;;;/p-3

InChI key

KOECRLKKXSXCPB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirko Scholz et al.
Physical chemistry chemical physics : PCCP, 20(16), 10677-10685 (2017-12-21)
Trivalent bismuth compounds featuring octahedral halide coordination, such as BiI3, double-perovskite structures and vacancy-ordered perovskite derivatives, have recently attracted considerable attention as potential environmentally benign thin film materials for solar light-harvesting and optoelectronic applications aiming to replace toxic lead-based perovskites.
Yang Chen et al.
Physical chemistry chemical physics : PCCP, 13(13), 5659-5667 (2011-02-12)
The solvothermal reactions of BiI(3), KI, I(2), 4,4'-bipyridine (4,4'-bipy), and a small amount of water in alcohol and acetonitrile produced four bipyridinium iodobismuthates {[MQ](3)[Bi(2)I(6)(μ-I)(3)][Bi(2)I(6)(μ-I)(2)(MQ)(2)](3)} (1, MQ(+) = N-methyl-4,4'-bipyridinium), {[EQ](3)[Bi(2)I(6)(μ-I)(3)][Bi(2)I(6)(μ-I)(2)(EQ)(2)](3)} (2, EQ(+) = N-ethyl-4,4'-bipyridinium), [MV][BiI(5)] (Eur. J. Inorg. Chem., 2010, 5326)
Minhua Su et al.
Journal of hazardous materials, 229-230, 72-82 (2012-06-20)
BiOI-MWCNT composites, with high absorption and visible-light photocatalytic performance, were synthesized by a solvothermal process, in which ethylene glycol (EG) participated in the reaction. Synthesized BiOI-MWCNT composites were characterized by X-ray diffraction (XRD), diffuse reflectance spectra (DRS), scanning electronic microscopy
Bismuth iodide impregnation of the peripheral nerve fibres.
S Dolapchieva et al.
Zeitschrift fur mikroskopisch-anatomische Forschung, 99(5), 804-812 (1985-01-01)
F M Abdel-Gawad
Journal of pharmaceutical and biomedical analysis, 16(5), 793-799 (1998-04-16)
A systematic spectrophotometric study on the ion-pair formation of Bi(III)-iodide with amineptine hydrochloride, piribedil and trimebutine maleate is carried out. The optimal experimental conditions pH, concentration of Bi(III) nitrate, potassium iodide; and the nature and amount of organic solvent have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service