Skip to Content
Merck
All Photos(2)

Documents

229237

Sigma-Aldrich

4,4′-Dibromobiphenyl

98%

Synonym(s):

1-Bromo-4-(4-bromophenyl)benzene, 4,4′-Dibromo-1,1′-biphenyl, 4,4′-Dibromodiphenyl, 4′,4-Dibromobiphenyl, PBB 15, p,p′-Dibromobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4C6H4Br
CAS Number:
Molecular Weight:
312.00
Beilstein:
2044701
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

355-360 °C (lit.)

mp

163-165 °C (lit.)

SMILES string

Brc1ccc(cc1)-c2ccc(Br)cc2

InChI

1S/C12H8Br2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H

InChI key

HQJQYILBCQPYBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The degradation of 4,4′-dibromobiphenyl, through photoelectrocatalytic process, was studied.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanshan Qiu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(5), 429-434 (2010-05-11)
In this work, both experimental and theoretical study on the FT-IR and Raman spectra as well as (1)H NMR and (13)C NMR chemical shifts of 4,4'-dibromodiphenyl ether have been carried out. The optimized geometry was obtained by using both HF
Haijin Liu et al.
Chemosphere, 82(1), 43-47 (2010-11-03)
Polybrominated biphenyls have been widely used as flameretardants in textile and electronic industries, and additives in plastics. Over the past few decades, much attention has been given to polybrominated biphenyls in the environment and their effects on humans. In this
Marola M H van Lipzig et al.
Chemical research in toxicology, 18(11), 1691-1700 (2005-11-23)
Exposure of humans and wildlife to xenobiotics, such as halogenated biphenyls, that interfere with the endogenous estrogen balance may lead to endocrine disruption. Such compounds may either mimic or block estradiol's action by agonistic or antagonistic action, respectively. They may
Li Mao et al.
Huan jing ke xue= Huanjing kexue, 31(1), 199-204 (2010-03-25)
The effect of copper ions on the water solubility enhancement of 4,4'- dibromodiphenyl ethers (BDE-15) by a commercial humic acid (HA) has been studied using batch equilibrium technique at various pHs. The characteristics of HA-Cu complex were investigated with the
Aurélien Rey et al.
Journal of chromatography. A, 1404, 95-103 (2015-06-09)
A new strategy was developed to elucidate and quantify 56 (69 analytes including isomers) suspected chemically defined fragrance allergens in perfumes that were recently targeted by the Scientific Committee on Consumer Safety (SCCS). Samples were analyzed with a two-dimensional gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service