Skip to Content
Merck
All Photos(3)

Documents

223735

Sigma-Aldrich

Coniferyl alcohol

98%

Synonym(s):

3-(4-Hydroxy-3-methoxyphenyl)-2-propen-1-ol, 4-Hydroxy-3-methoxycinnamyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH=CHCH2OH
CAS Number:
Molecular Weight:
180.20
Beilstein:
2048961
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

163-165 °C/3 mmHg (lit.)

mp

75-80 °C (lit.)

solubility

alcohol: moderately soluble(lit.)
diethyl ether: freely soluble(lit.)
water: insoluble (almost)

storage temp.

−20°C

SMILES string

COc1cc(\C=C\CO)ccc1O

InChI

1S/C10H12O3/c1-13-10-7-8(3-2-6-11)4-5-9(10)12/h2-5,7,11-12H,6H2,1H3/b3-2+

InChI key

JMFRWRFFLBVWSI-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Coniferyl alcohol is one of the preferred substrate of the eucalpytus globus enzyme.

Application

Coniferyl alcohol was used as a fungal growth inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Renán García et al.
Plant cell reports, 33(8), 1263-1274 (2014-04-17)
The gene coding for F5H from Eucalyptus globulus was cloned and used to transform an f5h -mutant of Arabidopsis thaliana , which was complemented, thus verifying the identity of the cloned gene. Coniferaldehyde 5-hydroxylase (F5H; EC 1.14.13) is a cytochrome
Stefanie König et al.
The New phytologist, 202(3), 823-837 (2014-02-04)
Verticillium longisporum is a soil-borne vascular pathogen causing economic loss in rape. Using the model plant Arabidopsis this study analyzed metabolic changes upon fungal infection in order to identify possible defense strategies of Brassicaceae against this fungus. Metabolite fingerprinting identified
Paula Oyarce et al.
Nature plants, 5(2), 225-237 (2019-01-30)
Lignin is the main cause of lignocellulosic biomass recalcitrance to industrial enzymatic hydrolysis. By partially replacing the traditional lignin monomers by alternative ones, lignin extractability can be enhanced. To design a lignin that is easier to degrade under alkaline conditions
Guilhem Reyt et al.
Nature communications, 12(1), 2320-2320 (2021-04-21)
Lignin is a complex polymer deposited in the cell wall of specialised plant cells, where it provides essential cellular functions. Plants coordinate timing, location, abundance and composition of lignin deposition in response to endogenous and exogenous cues. In roots, a
Alif Meem Nurani et al.
Plant & cell physiology, 61(2), 255-264 (2020-01-11)
Stem cells undergo cell division and differentiation to ensure organized tissue development. Because plant cells are immobile, plant stem cells ought to decide their cell fate prior to differentiation, to locate specialized cells in the correct position. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service