Skip to Content
Merck
All Photos(1)

Documents

223034

Sigma-Aldrich

1-Phenyl-1,2-propanedione

99%

Synonym(s):

Acetyl benzoyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCOCH3
CAS Number:
Molecular Weight:
148.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

impurities

1% methyl benzoate

refractive index

n20/D 1.532 (lit.)

bp

103-105 °C/14 mmHg (lit.)

density

1.101 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C(=O)c1ccccc1

InChI

1S/C9H8O2/c1-7(10)9(11)8-5-3-2-4-6-8/h2-6H,1H3

InChI key

BVQVLAIMHVDZEL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The enantioselective hydrogenation of 1-phenyl-1,2-propanedione over Pt colloids was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Cunha Brandt et al.
Journal of dentistry, 39(6), 438-447 (2011-04-23)
The aim of this study was to evaluate the degree of conversion (DC), rate of polymerization (R(p)(max)), Knoop hardness (KHN) and bond strength between tooth/restoration of composite resins containing different photo-initiators photo-activated by different light-curing units (LCUs). A mixture of
Luis Felipe J Schneider et al.
Dental materials : official publication of the Academy of Dental Materials, 25(3), 369-375 (2008-10-14)
To evaluate the effect of amine ratio (ethyl 4-dimethylaminobenzoate, EDMAB) on the maximum rate of polymerization (R(p)(max)), degree of conversion (DC), Knoop hardness (KH), water sorption (Wsp), water solubility (Wsl) and color changes (DeltaE) over time of resin composites formulated
Hiroyuki Arikawa et al.
Dental materials journal, 28(4), 454-460 (2009-09-02)
The purpose of this study was to investigate effects of various visible light photoinitiators on the polymerization efficiency and color of the light-activated resins. Four photoinitiators, including camphorquinone, phenylpropanedione, monoacrylphosphine oxide (TPO), and bisacrylphosphine oxide (Ir819), were used. Each photoinitiator
Marcello G Trevisan et al.
Talanta, 75(4), 1021-1027 (2008-07-01)
Fourier transform mid-infrared spectroscopy (FT-MIR) coupled with a homemade attenuated total reflectance (ATR) flow-cell was used for on-line monitoring of a biotransformation reaction. The reaction was also monitored off-line by gas chromatography-mass spectrometry (GC-MS) enabling to establish a multivariate model
Miguel G Neumann et al.
Dental materials : official publication of the Academy of Dental Materials, 22(6), 576-584 (2005-11-18)
To evaluate the efficiency of the photopolymerization of dental resins it is necessary to know to what extent the light emitted by the light curing units is absorbed by the photoinitiators. On the other hand, the efficiency of the absorbed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service