Skip to Content
Merck
All Photos(1)

Key Documents

22134

Sigma-Aldrich

(+)-β-Cedrene

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(1S,2R,5S)-8-Methylene-2,6,6-trimethyltricyclo[5.3.1.01.5]undecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2244722
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]/D 11.5 to 14.5°, c = 1 in chloroform

refractive index

n20/D 1.502

bp

263-264 °C (lit.)

density

0.932 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]1(C2=C)C(C)(C)[C@@](CC[C@H]3C)([H])[C@]3(CC2)C1

InChI

1S/C15H24/c1-10-7-8-15-9-12(10)14(3,4)13(15)6-5-11(15)2/h11-13H,1,5-9H2,2-4H3/t11-,12+,13+,15+/m1/s1

InChI key

DYLPEFGBWGEFBB-OSFYFWSMSA-N

General description

β-Cedrene is a tricyclic sesquiterpene obtained from Juniperus cedrus and Juniperus thurifera. It is the main constituent of cedarwood oil, which is used as a raw material for biofuel development.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of a highly effective intramolecular Pauson-Khand cyclisation for the formal total synthesis of (?)-α-and β-cedrene by preparation of cedrone
Crawford JJ, et al.
Tetrahedron, 62(49), 11360-11370 (2006)
Renewable high density fuels containing tricyclic sesquiterpanes and alkyl diamondoids
Harrison KW and Harvey BG
Sustainable Energy & Fuels, 1(3), 467-473 (2017)
Formal Total Synthesis of (?)-α-and β-Cedrene by Preparation of Cedrone. Construction of the Tricyclic Carbon Skeleton by the Use of a Highly Efficient Intramolecular Khand Annulation
Kerr WJ, et al.
Organic Letters, 3(19), 2945-2948 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service