Skip to Content
Merck
All Photos(1)

Key Documents

218006

Sigma-Aldrich

5-Chloro-3-hydroxypyridine

99%

Synonym(s):

5-Chloro-3-pyridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4ClNO
CAS Number:
Molecular Weight:
129.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

160-162 °C (lit.)

SMILES string

Oc1cncc(Cl)c1

InChI

1S/C5H4ClNO/c6-4-1-5(8)3-7-2-4/h1-3,8H

InChI key

TUIDQYRZDZRHPQ-UHFFFAOYSA-N

General description

5-Chloro-3-hydroxypyridine undergoes Suzuki-Miyaura cross-coupling with 3-pyridylboronic acid in the presence of Pd catalyst to yield hydroxybiphenyls.

Application

5-Chloro-3-hydroxypyridine was used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robbins TA, et al.
Journal of the American Chemical Society, 116, 111-111 (1994)
Microwave-promoted Suzuki-Miyaura coupling of arylboronic acids with 1-bromo-2-naphthol, o-bromophenol, and o-chlorophenol.
Wawrzyniak P and Heinicke J.
Tetrahedron Letters, 47(50), 8921-8924 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service