Skip to Content
Merck
All Photos(4)

Key Documents

215813

Sigma-Aldrich

Potassium hydride

greener alternative

30 wt % dispersion in mineral oil

Synonym(s):

Potassium monohydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH
CAS Number:
Molecular Weight:
40.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

slurry

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

30 wt % dispersion in mineral oil

greener alternative category

SMILES string

[K]

InChI

1S/K.H

InChI key

NTTOTNSKUYCDAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patnaik P et al.
A Comprehensive Guide to the Hazardous Properties of Chemical Substances null
Potassium hydride in paraffin: A useful base for organic synthesis.
Taber DF and Nelson CG
The Journal of Organic Chemistry, 71(23), 8973-8974 (2006)
Pieter Clauw et al.
Plant physiology, 167(3), 800-816 (2015-01-22)
Although the response of plants exposed to severe drought stress has been studied extensively, little is known about how plants adapt their growth under mild drought stress conditions. Here, we analyzed the leaf and rosette growth response of six Arabidopsis
Stefan D Anker et al.
European journal of heart failure, 17(10), 1050-1056 (2015-05-27)
Hyperkalaemia in heart failure patients limits use of cardioprotective renin-angiotensin-aldosterone system inhibitors (RAASi). Sodium zirconium cyclosilicate (ZS-9) is a selective potassium ion trap, whose mechanism of action may allow for potassium binding in the upper gastrointestinal tract as early as
Jordan E Bisanz et al.
Applied and environmental microbiology, 81(15), 4965-4975 (2015-05-17)
The nutritional status of pregnant women is vital for healthy outcomes and is a concern for a large proportion of the world's population. The role of the microbiota in pregnancy and nutrition is a promising new area of study with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service