Skip to Content
Merck
All Photos(1)

Key Documents

215767

Sigma-Aldrich

Disperse Red 19

Dye content 97 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O4
CAS Number:
Molecular Weight:
330.34
Colour Index Number:
11130
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

composition

Dye content, 97%

mp

>300 °C (lit.)

λmax

495 nm

SMILES string

OCCN(CCO)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C16H18N4O4/c21-11-9-19(10-12-22)15-5-1-13(2-6-15)17-18-14-3-7-16(8-4-14)20(23)24/h1-8,21-22H,9-12H2/b18-17+

InChI key

GHDZRIQTRDZCMV-ISLYRVAYSA-N

General description

Disperse Red 19 (DR19) is widely used as a non-linear optical (NLO) dye with two hydroxyl (OH) groups, which can covalently connect with the base polymer. It can be linked as a side chain group that provides thermal and photo stability to the polymeric unit.

Application

DR19 can be used as red side groups in an NLO polyester for potential application in optical switching devices. It can also be bonded with polyurethane to form stable films, which may be used in the fabrication of analog systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nonlinear optics and optical micropatterning in polymers with disperse red 19 sidegroups
Nonlinear Optical Properties of Organic Materials V, 1775, 379-391 (1993)
New third-order nonlinear polymers functionalized with disperse red and disperse orange chromophores with increased stability
Cherioux, F., Audebert, P., Maillotte, H., Grossard, L., Hernandez, F. E., & Lacourt, A.
Chemistry of Materials, 9(12), 2921-2927 (1997)
Diacetylene-containing polymers. XIII. Poly (hexa-2, 4-diynylene-l, 6-dioxydicinnamates) containing an azo dye, Disperse Red 19
Miura H and Ogawa T
Polymer Bull., 49(2-3), 103-110 (2002)
Thermosetting nonlinear optical polymer: Polyurethane with disperse red 19 side groups
Shi Y, et al.
Applied Physics Letters, 60(21), 2577-2579 (1992)
Large photoinduced birefringence in an optically nonlinear polyester polymer
Shi Y, et al.
Applied Physics Letters, 59(23), 2935-2937 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service