Skip to Content
Merck
All Photos(2)

Documents

204889

Sigma-Aldrich

Ytterbium(III) oxide

99.99% trace metals basis

Synonym(s):

Ytterbium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb2O3
CAS Number:
Molecular Weight:
394.08
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38161601
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: ytterbium

SMILES string

O=[Yb]O[Yb]=O

InChI

1S/3O.2Yb

InChI key

FIXNOXLJNSSSLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Multifunctional Lanthanide-Based Metal-Organic Frameworks with a Polyheterotopic Ligand: Doped with Ytterbium(III) for Luminescence Enhancement and Selective Dye Adsorption.: This study investigates the application of ytterbium(III) oxide doped in metal-organic frameworks (MOFs) to enhance luminescence and selectively adsorb dyes, highlighting its potential in advanced energy storage and battery technology. The multifunctionality of these materials opens up new possibilities for energy-efficient solutions in battery development (Chen et al., 2018).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Heublein et al.
Journal of dairy science, 100(2), 1019-1036 (2016-12-13)
The objective of this study was to test whether diet selection of dairy cows under grazing conditions could be estimated using plant wax markers. Furthermore, differences between 2 cow strains and the effect of concentrate supplementation on plant species selection
Zhengze Yu et al.
ACS nano, 9(11), 11064-11074 (2015-10-13)
Photodynamic therapy (PDT) is a well-established modality for cancer therapy, which locally kills cancer cells when light irradiates a photosensitizer. However, conventional PDT is often limited by the extremely short lifespan and severely limited diffusion distance of reactive oxygen species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service