All Photos(2)
About This Item
Empirical Formula (Hill Notation):
HfO2
CAS Number:
Molecular Weight:
210.49
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
≥99.95%
form
powder
density
9.68 g/mL at 25 °C (lit.)
SMILES string
O=[Hf]=O
InChI
1S/Hf.2O
InChI key
CJNBYAVZURUTKZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service