Skip to Content
Merck
All Photos(1)

Key Documents

202657

Sigma-Aldrich

Arsenic

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As
CAS Number:
Molecular Weight:
74.92
EC Number:
MDL number:
UNSPSC Code:
12141906
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

resistivity

33.3 μΩ-cm

bp

613 °C (lit.)

mp

817 °C (lit.)

density

5.727 g/mL at 25 °C (lit.)

SMILES string

[As]

InChI

1S/As

InChI key

RQNWIZPPADIBDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Arsenic pieces may be used to prepare α-realgar (As4S4).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Repr. 1A - STOT RE 1 Oral

Target Organs

Respiratory organs,Cardio-vascular system,Gastro-intestinal system,Central nervous system,Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valence and core-level binding energy shifts in realgar (As 4 S 4) and pararealgar (As 4 S 4) arsenic sulfides.
Bullen HA, et al.
Surface Science, 531(3), 319-328 (2003)
Meryene C Teixeira et al.
Food chemistry, 154, 38-43 (2014-02-13)
We have developed an eletroanalytical method that employs Cu(2+) solutions to determine arsenic in sugarcane brandy using an electrode consisting of carbon paste modified with carbon nanotubes (CNTPE) and polymeric resins. We used linear sweep (LSV) and differential-pulse (DPV) voltammetry
Ilona Dudka et al.
Environment international, 68, 71-81 (2014-04-10)
Environmental metabonomics is the application of metabonomics to characterize the interactions of organisms with their environment. Metabolic profiling is an exciting addition to the armory of the epidemiologist for the discovery of new disease risk biomarkers and diagnostics. This work
Arsenic binding to proteins.
Shengwen Shen et al.
Chemical reviews, 113(10), 7769-7792 (2013-07-03)
Lucia Cavalca et al.
Future microbiology, 8(6), 753-768 (2013-04-17)
Arsenic is present in many environments and is released by various natural processes and anthropogenic actions. Although arsenic is recognized to cause a wide range of adverse health effects in humans, diverse bacteria can metabolize it by detoxification and energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service