Skip to Content
Merck
All Photos(1)

Key Documents

192708

Sigma-Aldrich

Triethylaluminum solution

25 wt. % in toluene

Synonym(s):

Aluminumtriethanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(C2H5)3
CAS Number:
Molecular Weight:
114.16
Beilstein:
3587229
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

25 wt. % in toluene

density

0.848 g/mL at 25 °C

SMILES string

CC[Al](CC)CC

InChI

1S/3C2H5.Al/c3*1-2;/h3*1H2,2H3;

InChI key

VOITXYVAKOUIBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylaluminumsolution (triethylaluminum in toluene solution), is commonly used as a catalyst in organic synthesis.

Application

Triethylaluminum solution is used as a catalyst in the preparation of (2-chloroethyl)oxirane,(3-chloropropyl)oxirane, and (4-chlorobutyl)oxirane from the corresponding alkenols.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Self-heat. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Poly [(2-chloroethyl) oxirane], poly [(3-chloropropyl)-oxirane] and poly [(4-chlorobutyl) oxirane]: New polyether elastomers of improved chemical reactivity
Shih JS, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 20(10), 2839-2849 (1982)
Aliphatic polyester-based biodegradable materials: new amphiphilic graft copolymers
Rutot D, et al.
Polymer Degradation and Stability, 73(3), 561-566 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service