Skip to Content
Merck
All Photos(2)

Key Documents

190705

Sigma-Aldrich

Ethyl 4-chloro-4-oxobutyrate

94%

Synonym(s):

Ethyl succinyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCH2CH2CO2C2H5
CAS Number:
Molecular Weight:
164.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

94%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

88-90 °C/11 mmHg (lit.)

density

1.155 g/mL at 25 °C (lit.)

functional group

acyl chloride
ester

storage temp.

2-8°C

SMILES string

CCOC(=O)CCC(Cl)=O

InChI

1S/C6H9ClO3/c1-2-10-6(9)4-3-5(7)8/h2-4H2,1H3

InChI key

IXZFDJXHLQQSGQ-UHFFFAOYSA-N

Related Categories

Application

Ethyl 4-chloro-4-oxobutyrate was used in the synthesis of:
  • polymer-supported bifunctional catalyst
  • succinimidyl ester of benzo(a)pyrene
  • succinate prodrugs of curcuminoids

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wisut Wichitnithad et al.
Molecules (Basel, Switzerland), 16(2), 1888-1900 (2011-02-24)
A novel series of succinyl derivatives of three curcuminoids were synthesized as potential prodrugs. Symmetrical (curcumin and bisdesmethoxycurcumin) and unsymmetrical (desmethoxycurcumin) curcuminoids were prepared through aldol condensation of 2,4-pentanedione with different benzaldehydes. Esterification of these compounds with a methyl or
Surface plasmon resonance detection of endocrine disruptors using immunoprobes based on self-assembled monolayers.
Gobi KV, et al.
Sensors and Actuators B, Chemical, 108(1), 784-790 (2005)
Enantioselective Strecker-type reaction promoted by polymer-supported bifunctional catalyst.
Nogami H, et al.
Tetrahedron Letters, 42(2), 279-283 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service