All Photos(1)
About This Item
Linear Formula:
ClCH2COOC(CH3)3
CAS Number:
Molecular Weight:
150.60
Beilstein:
1753006
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.423 (lit.)
bp
48-49 °C/11 mmHg (lit.)
density
1.053 g/mL at 25 °C (lit.)
functional group
chloro
ester
SMILES string
CC(C)(C)OC(=O)CCl
InChI
1S/C6H11ClO2/c1-6(2,3)9-5(8)4-7/h4H2,1-3H3
InChI key
KUYMVWXKHQSIAS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
tert-Butyl chloroacetate was used in the synthesis of:
- imidazol-1-yl-acetic acid hydrochloride
- cis-disubstituted aziridine ester via aza-Darzens reaction
- 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
WGK
WGK 1
Flash Point(F)
114.8 °F
Flash Point(C)
46 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
E Vedejs et al.
The Journal of organic chemistry, 65(18), 5498-5505 (2000-09-02)
Intermolecular alkylation of the aziridinyl oxazole 20 using PhSO(2)CH(2)CH(2)OTf is possible despite the presence of potentially nucleophilic aziridine nitrogen. The resulting oxazolium salt 22 reacts with BnNMe(3)(+)CN(-) to produce the azomethine ylide 24b via electrocyclic ring opening of an oxazoline
Santosh Kumar Singh et al.
Beilstein journal of organic chemistry, 4, 42-42 (2008-12-24)
A convenient and practical synthesis of imidazol-1-yl-acetic acid hydrochloride was achieved via N-alkylation of imidazole using tert-butyl chloroacetate followed by a non-aqueous ester cleavage of the resulting imidazol-1-yl-acetic acid tert-butyl ester in the presence of titanium tetrachloride. The synthesized imidazol-1-yl-acetic
Substituent dependent fluorescence response of diazacrown-based PET sensors.
Nagy K, et al.
Tetrahedron, 64(27), 6191-6195 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service