Skip to Content
Merck
All Photos(1)

Documents

183644

Sigma-Aldrich

2-Bromonaphthalene

97%

Synonym(s):

β-Bromonaphthalene, β-Naphthyl bromide, 2-Naphthyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Br
CAS Number:
Molecular Weight:
207.07
Beilstein:
1858110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

impurities

≤3% 1-bromonaphthalene

bp

281-282 °C (lit.)

mp

52-55 °C (lit.)

solubility

methanol: soluble 50 mg/mL, clear, colorless to yellow

SMILES string

Brc1ccc2ccccc2c1

InChI

1S/C10H7Br/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

APSMUYYLXZULMS-UHFFFAOYSA-N

Gene Information

human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 2-bromonaphthalene with cuprous cyanide in N-methylpyrrolidone has been investigated. Nanosecond time-resolved resonance Raman spectra of the T1→Tn transition of 2-bromonaphthalene in methanol solvent has been investigated.

Application

2-Bromonaphthalene was used in the synthesis of biaryls via Suzuki cross coupling reaction in water under microwave irradiation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid microwave-promoted Suzuki cross coupling reaction in water.
Bai L, et al.
Green Chemistry, 5(5), 615-617 (2003)
Isaiah R Speight et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
Owing to the strength of the C-F bond, the 'direct' preparation of Grignard reagents, i.e., the interaction of elemental magnesium with an organic halide, typically in an ethereal solvent, fails for bulk magnesium and organofluorine compounds. Previously described mechanochemical methods
Notes-A Convenient Synthesis of Water-Soluble Carbodiimides.
Sheehan J, et al.
The Journal of Organic Chemistry, 26(7), 2525-2528 (1961)
Nanosecond time-resolved resonance Raman investigation of the T1→ Tn transition of 2-bromonaphthalene.
Shoute LCT, et al.
Chemical Physics Letters, 290(1), 24-28 (1998)
J M Shoulder et al.
SAR and QSAR in environmental research, 30(4), 229-245 (2019-03-22)
Persistent organic contaminants in the environment pose an environmental risk due to widespread occurrence and toxic properties. Advanced oxidation processes (AOPs) are treatment methods that have been used to successfully degrade organic contaminants in water, soil, sediments and sludge. Reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service