166375
Formic hydrazide
Synonym(s):
Formic acid hydrazide, Formylhydrazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HCONHNH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635759
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
mp
52-57 °C (lit.)
solubility
water: soluble 2.5%, clear, colorless
functional group
amine
hydrazine
SMILES string
[H]C(=O)NN
InChI
1S/CH4N2O/c2-3-1-4/h1H,2H2,(H,3,4)
InChI key
XZBIXDPGRMLSTC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Formic hydrazide was used in the synthesis of 1,2,4-triazole derivatives. It was also used in the synthesis of new anticancer agent, 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(β-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
233.6 °F - closed cup
Flash Point(C)
112 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Traceless liquid-phase synthesis of 3-alkylamino-4, 5-disubstituted-1, 2, 4-triazoles on polyethylene glycol (PEG).
Zong Y-X, et al.
Tetrahedron Letters, 46(31), 5139-5141 (2005)
B Toth et al.
Carcinogenesis, 1(1), 61-65 (1980-01-01)
N-ethyl-N-formylhydrazine (EFH) was administered as a 0.02% solution in drinking water continuously for life to randomly bred Swiss mice, from 6 weeks of age. The treatment induced tumors of the lungs, blood vessels, liver, gall bladder and preputial glands. The
Synthesis of NB-506, a new anticancer agent.
Ohkubo M, et al.
Tetrahedron, 53(2), 585-592 (1997)
Coro Echeverría et al.
Polymers, 11(6) (2019-06-05)
Nano-sized one-dimensional metallo-organic polymers, characterized by the phenomenon of spin transition, are excellent candidates for advanced technological applications such as optical sensors, storage, and information processing devices. However, the main drawback of this type of polymers is their fragile mechanical
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service