Skip to Content
Merck
All Photos(3)

Key Documents

163511

Sigma-Aldrich

Hydroxymethanesulfinic acid monosodium salt dihydrate

Synonym(s):

Sodium formaldehydesulfoxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2SO2Na · 2H2O
CAS Number:
Molecular Weight:
154.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

64-68 °C (lit.)

solubility

alcohol: slightly soluble(lit.)
benzene: slightly soluble(lit.)
chloroform: slightly soluble(lit.)
diethyl ether: slightly soluble(lit.)
water: freely soluble(lit.)

functional group

sulfinic acid

SMILES string

[Na+].[H]O[H].[H]O[H].OCS([O-])=O

InChI

1S/CH4O3S.Na.2H2O/c2-1-5(3)4;;;/h2H,1H2,(H,3,4);;2*1H2/q;+1;;/p-1

InChI key

UCWBKJOCRGQBNW-UHFFFAOYSA-M

General description

Oxidation reation of hydroxymethanesulfinic acid with chlorite has been studied in the pH range 4-8. It is a sulfur-containing reducing agent.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxyhalogen-Sulfur Chemistry: Oxidation of Hydroxymethanesulfinic Acid by Chlorite 1.
Salem MA, et al.
The Journal of Physical Chemistry, 100(22), 9377-9384 (1996)
Recent trends in the chemistry of sulfur-containing reducing agents.
Makarov, SV.
Russ. Chem. Rev., 70(10), 885-895 (2001)
S V Makarov et al.
Archives of biochemistry and biophysics, 367(2), 289-296 (1999-07-09)
Sodium hydroxymethanesulfinate, (HOCH2SO2Na, HMS) is relatively stable in aqueous alkaline environments, but rapidly decomposes in acidic medium to give a variety of products that include sulfur dioxide. A detailed kinetic and mechanistic study of the decomposition of HMS in slightly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service