Skip to Content
Merck
All Photos(1)

Key Documents

163015

Sigma-Aldrich

Thallium(III) nitrate trihydrate

Synonym(s):

TTN, Thallium trinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tl(NO3)3 · 3H2O
CAS Number:
Molecular Weight:
444.44
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: thallium
reagent type: catalyst

mp

102-105 °C (lit.)

SMILES string

O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.3H2O.Tl/c3*2-1(3)4;;;;/h;;;3*1H2;/q3*-1;;;;+3

InChI key

ZCPIKMRXMBJLCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the rapid oxidation of styrene derivatives to carbonyl compounds, for the regioselective ring expansion of cyclic aralkyl ketones via oxidative rearrangement, for the selective hydrolysis of thioacetals to carbonyl compounds and for a general synthetic method to 19-norsteroids. Mediates ring contraction of cyclic ketones to a series of trans-fused 10-methyl-2-decalones.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 9, 301-301 (1979)
Kevin Yauy et al.
The Journal of molecular diagnostics : JMD, 20(4), 465-473 (2018-04-25)
Interpretation of next-generation sequencing constitutes the main limitation of molecular diagnostics. In diagnosing myopathies and muscular dystrophies, another issue is efficiency in predicting the pathogenicity of variants identified in large genes, especially TTN; current in silico prediction tools show limitations
Enrico Bugiardini et al.
Frontiers in neurology, 9, 456-456 (2018-07-13)
Objective: Distal myopathies are a diagnostically challenging group of diseases. We wanted to understand the value of MRI in the current clinical setting and explore the potential for optimizing its clinical application. Methods: We retrospectively audited the diagnostic workup in
Sophie Lalande et al.
Experimental physiology, 102(9), 1055-1066 (2017-08-02)
What is the topic of this review? This review focuses on how in vivo and molecular measurements of cardiac passive stiffness can predict exercise tolerance and how exercise training can reduce cardiac passive stiffness. What advances does it highlight? This
Journal of the Chemical Society. Chemical Communications, 1026-1026 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service