All Photos(3)
About This Item
Linear Formula:
CH3C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
94.54
Beilstein:
3591762
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
mp
165-170 °C (lit.)
functional group
amidine
SMILES string
Cl[H].CC(N)=N
InChI
1S/C2H6N2.ClH/c1-2(3)4;/h1H3,(H3,3,4);1H
InChI key
WCQOBLXWLRDEQA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Acetamidine hydrochloride is an amidine salt and its conversion to 2,4,6-trimethyl-sym-triazine has been studied.
Application
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
H Niwa et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(24), 13617-13622 (1996-11-26)
The jellyfish Aequorea victoria possesses in the margin of its umbrella a green fluorescent protein (GFP, 27 kDa) that serves as the ultimate light emitter in the bioluminescence reaction of the animal. The protein is made up of 238 amino
Michael Schnoor et al.
Biochemical and biophysical research communications, 322(3), 867-872 (2004-09-01)
Different substances such as dimethyl sulfoxide, tetramethylene sulfoxide, 2-pyrollidone, and the naturally occurring compatible solute betaine enhance PCR amplification of GC-rich DNA templates with high melting temperatures. In particular, cyclic compatible solutes outperform traditional PCR enhancers. We therefore investigated the
Cristina Maccallini et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 8(6), 991-995 (2012-06-30)
A new series of phenyl- and heteryl acetamidines were synthesized and evaluated as inhibitors of nitric oxide synthases (NOS). While the N-substitution of the acetamidine moiety with different heterocycles appears to completely destroy the activity, linking the phenyl core preserves
Justin E Jones et al.
Bioorganic chemistry, 38(2), 62-73 (2009-12-29)
Helicobacter pylori encodes a potential virulence factor, agmatine deiminase (HpAgD), which catalyzes the conversion of agmatine to N-carbamoyl putrescine (NCP) and ammonia - agmatine is decarboxylated arginine. Agmatine is an endogenous human cell signaling molecule that triggers the innate immune
Jin Sheng Guo et al.
European journal of pharmacology, 536(3), 301-308 (2006-04-08)
Nitric oxide synthases (NOS) and cyclooxygenase-2 (COX-2) are important enzymes involved in ulcer healing but interactions between them have not been clearly defined. The aim of this study was to investigate the effects of selective or non-selective inhibition of NOS
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service