Skip to Content
Merck
All Photos(3)

Documents

156124

Sigma-Aldrich

Oxonic acid potassium salt

97%

Synonym(s):

4,6-Dihydroxy-1,3,5-triazine-2-carboxylic acid potassium salt, Allantoxanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2KN3O4
CAS Number:
Molecular Weight:
195.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

300 °C (lit.)

SMILES string

[K+].[O-]C(=O)C1=NC(=O)NC(=O)N1

InChI

1S/C4H3N3O4.K/c8-2(9)1-5-3(10)7-4(11)6-1;/h(H,8,9)(H2,5,6,7,10,11);/q;+1/p-1

InChI key

IAPCTXZQXAVYNG-UHFFFAOYSA-M

Application

Oxonic acid potassium salt is an inhibitor of uricase. The product from Sigma has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieyu Zuo et al.
The Journal of pharmacy and pharmacology, 70(1), 124-132 (2017-10-24)
Traditional Chinese herbal formulas are difficult to be understood because of complex compositions and specific therapeutic principles. To better understand herbal compatibility in Traditional Chinese medicine (TCM), this study was conducted to investigate the effects of a Chinese pharmacopoeia-listed formula
T Shirasaka et al.
Cancer research, 53(17), 4004-4009 (1993-09-01)
The possibility of decreasing the gastrointestinal (GI) toxic effects of 5-fluorouracil (5-FU) on the digestive tract such as its injury of cells and induction of diarrhea, without reducing its antitumor activity, was investigated in rats. Oxonic acid was found to
Kei Nishizawa et al.
PloS one, 14(4), e0214862-e0214862 (2019-04-06)
The effects of hyperuricemia on the expression of kidney drug transporters and on the pharmacokinetics of several substrate drugs were examined. We first established a rat model of hyperuricemia without marked symptoms of chronic kidney failure by 10-day co-administration of
Haruka Fujikawa et al.
Antioxidants (Basel, Switzerland), 9(5) (2020-05-10)
The oxidant/antioxidant imbalance plays a pivotal role in the lung. Uric acid (UA), an endogenous antioxidant, is highly present in lung tissue, however, its impact on lung function under pathophysiological conditions remains unknown. In this work, pharmacological and genetic inhibition
Yasushi Habu et al.
Biochemical pharmacology, 69(6), 993-999 (2005-03-08)
We previously reported that in hyperuricemic rats, renal impairment occurred and organic ion transport activity decreased, accompanied with a specific decrease in the expression of rat organic anion transporters, rOAT1 and rOAT3, and organic cation transporter, rOCT2. In the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service