Skip to Content
Merck
All Photos(1)

Key Documents

14501

Sigma-Aldrich

Poly(ethylene glycol) bis(amine)

Mw 2,000, carboxyl reactive, amine

Synonym(s):

Polyethylene glycol, O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) bis(amine), Mw 2,000

form

powder

mol wt

Mw 2,000

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

Ω-end

amine

α-end

amine

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Polyethylene Glycol Conjugated Polymeric Nanocapsules for Targeted Delivery of Quercetin to Folate-Expressing Cancer Cells in Vitro and in Vivo: Discusses the use of PEG-bis amine for creating polymeric nanocapsules aimed at enhancing targeted drug delivery (RI El-Gogary et al., 2014).

Other Notes

Polymer for preparing enzyme conjugates soluble in organic solvents; Promising drug carrier

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Kelemen et al.
Lab on a chip, 19(11), 1985-1990 (2019-05-03)
Whispering gallery mode (WGM) resonators are promising optical structures for microfluidic label-free biosensors mainly due to their high sensitivity, but from a practical point of view they present numerous constraints that make their use in real laboratory diagnosis application difficult.
K Kawasaki et al.
Chemical & pharmaceutical bulletin, 43(12), 2133-2138 (1995-12-01)
Hybrids of fibronectin-related peptides [Arg-Gly-Asp (RGD), Arg-Gly-Asp-Ser (RGDS)] and poly(ethylene glycol) (PEG) were prepared and their inhibitory effects on experimental metastasis in mice were examined. The inhibitory effect of RGD was markedly potentiated by hybrid formation with poly(ethylene glycol) #6000.
Urrotigoity, M. and Souppe, J.
Biocatalysis, 2, 145-145 (1989)
Stephen J Connon et al.
Bioorganic & medicinal chemistry letters, 12(14), 1873-1876 (2002-06-28)
The synthesis and olefin metathesis activity in protic solvents of 7, a phosphine-free ruthenium alkylidene bound to a hydrophilic solid support are reported. This heterogeneous catalyst promotes relatively efficient ring closing- and cross-metathesis reactions in both methanol and water. The
Gaëlle-Anne Cremer et al.
Journal of peptide science : an official publication of the European Peptide Society, 12(6), 437-442 (2006-01-25)
This paper describes the optimization of a synthesis of a difficult sequence related to a 12-mer sequence of a Pan DR epitope (PADRE). Elongation was followed by on-line monitoring of the N(alpha)-Fmoc removal adapted for the batch methodology. Studying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service